Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Heme oxygenase cleaves the heme ring at the alpha methane bridge to form biliverdin. Biliverdin is subsequently converted to bilirubin by biliverdin reductase. Under physiological conditions, the activity of heme oxygenase is highest in the spleen, where senescent erythrocytes are sequestrated and destroyed. Exhibits cytoprotective effects since excess of free heme sensitizes cells to undergo apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heat shock protein 32; heat shock protein, 32-kD; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decycling) 2; heme oxygenase (decyclizing) 1; heme oxygenase-2 (EC 1.14.99.3); hemoxygenase; HMOX1; HMOX2; HO-1; HO2; P32 protein; unnamed protein product
Gene Aliases: bK286B10; D8Wsu38e; Hemox; Heox; HEOXG; Hmox; HMOX1D; HO-1; HO-2; HO1; HO2; HSP32
UniProt ID: (Pig) P32394, (Human) P09601, (Chicken) P14791, (Human) P30519, (Rat) P06762, (Mouse) P14901, (Mouse) O70252, (Rat) P23711
Entrez Gene ID: (Pig) 445512, (Human) 3162, (Chicken) 396287, (Chicken) 416663, (Human) 3163, (Pig) 396622, (Rat) 24451, (Mouse) 15368, (Mouse) 15369, (Rat) 79239
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support