Invitrogen
PEP-100 is a 15 amino acid synthetic peptide whose sequence corresponds to amino acid residues 249-263 from human proteasome 20S C2 subunit. The sequence of this peptide is (amino to carboxy terminus): P (249)-A-D-E-P-A-E-K-A-D-E-P-M-E-H (263)
This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human proteasome 20S C2 subunit, catalog # PA1-963. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (~70-fold) for competitive inhibition of antibody-protein binding reactions.
Reconstitute with 0.1 mL of distilled water.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 30 kDa prosomal protein; epididymis secretory protein Li 275; Macropain subunit C2; macropain subunit nu; Multicatalytic endopeptidase complex subunit C2; PROS-30; proteasome (prosome, macropain) subunit, alpha type, 1; Proteasome component C2; Proteasome nu chain; Proteasome subunit alpha type-1; proteasome subunit nu; proteasome subunit, alpha-type, 1; protein P30-33K; testicular tissue protein Li 150
Gene Aliases: HC2; HEL-S-275; NU; PROS30; PSC2; PSMA1
UniProt ID: (Human) P25786
Entrez Gene ID: (Human) 5682
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support