Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
Assay-dependent | - |
Immunohistochemistry (IHC) |
Assay-dependent | - |
ELISA (ELISA) |
Assay-dependent | - |
Functional Assay (FN) |
Assay-dependent | View 5 publications 5 publications |
Control (Ctrl) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species |
Human |
Published species |
Human, Mouse |
Expression System |
HEK293 cells |
Molecular weight |
38 kDa |
Class |
Recombinant |
Type |
Protein |
Purity |
>95% by SDS-PAGE |
Endotoxin concentration |
<0.1 ng/µg |
Activity |
ED50 ≤ 5 ng/mL; determined by the dose-dependent proliferation of M-NSF60 cells. |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Purification |
sequential chromatography |
Contains |
no preservative |
Storage conditions |
4° C |
Reconstitution: Lyophilized human M-CSF should be reconstituted in 20mM Tris-HCl, pH 8.0 to a concentration of 0.1-1.0 mg/mL. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C.
Storage: Lyophilized human M-CSF should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted human M-CSF at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.
M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: colony stimulating factor 1 (macrophage); CSF-1; H-MCSF; Lanimostim; M-MCSF; macrophage colony stimulating factor 1; Macrophage colony-stimulating factor 1; MGC31930; Processed macrophage colony-stimulating factor 1
Gene Aliases: CSF-1; CSF1; MCSF
UniProt ID: (Human) P09603
Entrez Gene ID: (Human) 1435
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support