Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Histone H2A Antibodies

          Invitrogen

          Hydroxyl-Histone H2A (Tyr39) Recombinant Rabbit Monoclonal Antibody (K01_2Q23)

          View all (37) Histone H2A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Hydroxyl-Histone H2A (Tyr39) Recombinant Rabbit Monoclonal Antibody (K01_2Q23)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          Hydroxyl-Histone H2A (Tyr39) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Hydroxyl-Histone H2A (Tyr39) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Hydroxyl-Histone H2A (Tyr39) Antibody (MA564254) in IHC (P)

          Immunohistochemistry (Paraffin) analysis of Hydroxyl-Histone H2A (Tyr39) in paraffin-embedded Human lung cancer tissue. Samples were incubated with Hydroxyl-Histone H2A (Tyr39) recombinant monoclonal (Product # MA564254). High-pressure and temperature Sodium Citrate pH 6.0 was used for antigen retrieval. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Hydroxyl-Histone H2A (Tyr39) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Hydroxyl-Histone H2A (Tyr39) Antibody in Western Blot (WB)
          Hydroxyl-Histone H2A (Tyr39) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Hydroxyl-Histone H2A (Tyr39) Recombinant Rabbit Monoclonal Antibody (K01_2Q23)

          Product Details

          MA564254

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          K01_2Q23

          Immunogen

          A synthesized peptide derived from human Histone H2A (hydroxyl Y39).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A/G

          Storage buffer

          50mM tris glycine, pH 7.4, with 0.05% BSA, 150mM NaCl, 40% glycerol

          Contains

          0.01% sodium azide

          Storage conditions

          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3705302

          Target Information

          Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Acetyl-Histone H2; buforin I; H2A histone family member; H2A histone family member J; H2A histone family member V; H2A histone family member X; H2A histone family member Z; H2A histone family, member A; H2A histone family, member C; H2A histone family, member D; H2A histone family, member E; H2A histone family, member G; H2A histone family, member I; H2A histone family, member J; H2A histone family, member L; H2A histone family, member M; H2A histone family, member N; H2A histone family, member O; H2A histone family, member P; H2A histone family, member Q; H2A histone family, member R; H2A histone family, member V; H2A histone family, member X; H2A histone family, member Z; H2A.1; H2A.F/Z; H2A.W histone; H2A.Z variant histone 2; H2A/d; H2a/j; H2a/x; h2ad; H2ae; H2AFX; H2AFZ; h2ai; H2ak; H2AX histone; H2AZ histone; HIST1H3D; HIST2H2AA; HIST3H2A; histone (H2A.Z); histone 1, H2aa; histone 1, H2ab; histone 1, H2ac; histone 1, H2ad; histone 1, H2ae; histone 1, H2af; histone 1, H2ag; histone 1, H2ah; histone 1, H2ai; histone 1, H2aj; histone 1, H2ak; histone 1, H2al; histone 1, H2am; histone 1, H2an; histone 2, H2aa3; histone 2, H2aa4; histone 2, H2ab; histone 2, H2ac; histone 2, H2ai; Histone 2A; histone 3, H2a; histone 5 protein 2ax; histone cluster 1 H2A family member a; histone cluster 1 H2A family member b; histone cluster 1 H2A family member c; histone cluster 1 H2A family member d; histone cluster 1 H2A family member e; histone cluster 1 H2A family member g; histone cluster 1 H2A family member h; histone cluster 1 H2A family member i; histone cluster 1 H2A family member j; histone cluster 1 H2A family member k; histone cluster 1 H2A family member l; histone cluster 1 H2A family member m; histone cluster 1, H2aa; histone cluster 1, H2ab; histone cluster 1, H2ac; histone cluster 1, H2ad; histone cluster 1, H2ae; histone cluster 1, H2af; histone cluster 1, H2ag; histone cluster 1, H2ah; histone cluster 1, H2ai; histone cluster 1, H2ak; histone cluster 1, H2al; histone cluster 1, H2am; histone cluster 1, H2an; histone cluster 1, H2ao; histone cluster 1, H2ap; histone cluster 2 H2A family member a3; histone cluster 2 H2A family member a4; histone cluster 2 H2A family member b; histone cluster 2 H2A family member c; histone cluster 2, H2aa1; histone cluster 2, H2aa2; histone cluster 2, H2aa3; histone cluster 2, H2aa4; histone cluster 2, H2ab; histone cluster 2, H2ac; histone cluster 3 H2A; histone cluster 3, H2a; histone gene complex 2; histone H2a (aa 92-128); histone H2A type 1; Histone H2A type 1-B; Histone H2A type 1-E; Histone H2A type 1-G; Histone H2A type 1-I; histone H2A type 1-J; Histone H2A type 1-N; Histone H2A type 1-O; Histone H2A type 1-P; histone H2A-GL101; histone H2A.3; histone H2A.F/Z; histone H2A.x; histone H2A/a; histone H2A/g; histone H2A/l; histone H2A/m; histone H2A/p; histone H2A/ptl; histone H2A/q; histone H2A/r; histone H2A/s; histone H2AC; histone H2AD; histone H2AE; histone IIa; purine-rich binding element protein B; unnamed protein product

          View more View less

          Gene Aliases: 1190022L06; bA317E16.2; C530002L11Rik; dJ160A22.4; dJ193B12.1; dJ193B12.9; dJ221C16.4; dJ86C11.1; E130307C13; EG621893; gammaH2ax; Gm11276; H2A; H2a-221; H2a-291A; H2a-53; H2a-613a; H2a-613b; H2a-614; H2a-615; H2A-GL101; H2A.1; H2A.1b; H2A.2; H2A.3; H2A.i; H2A.X; H2A.Z; H2A.Z-1; H2A.Z-2; H2A.Z1; H2A.Z2; H2A/a; H2A/c; H2A/d; H2A/E; H2A/g; H2A/i; H2A/l; H2A/m; H2A/n; H2A/O; H2A/p; H2A/q; H2A/R; H2A/S; H2A/X; H2A/z; H2AA; H2AB; H2AFA; H2AFALii; H2AFC; H2AFD; H2AFE; H2AFG; H2AFI; H2AFJ; H2AFL; H2AFM; H2AFN; H2AFO; H2AFP; H2AFQ; H2AFR; H2AFV; H2AFX; H2AFZ; H2AG; H2AH; H2AV; H2AW; H2AZ; HIST1H2AA; HIST1H2AB; HIST1H2AC; HIST1H2AD; HIST1H2AE; Hist1h2af; HIST1H2AG; HIST1H2AH; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; Hist1h2an; Hist1h2ao; Hist1h2ap; Hist2; HIST2H2AA; Hist2h2aa1; Hist2h2aa2; HIST2H2AA3; HIST2H2AA4; HIST2H2AB; HIST2H2AC; HIST3H2A; Hist5-2ax; HISTH2AA; pH2A/f; Tg(Wnt1-cre)11Rth; TH2A; Wnt1-Cre; Wnt1::Cre; Wnt1cre

          View more View less

          UniProt ID: (Human) Q96QV6, (Human) P0C0S8, (Human) Q96KK5, (Human) Q99878, (Human) Q6FI13, (Human) Q16777, (Human) Q8IUE6, (Human) Q7L7L0, (Human) P04908, (Human) Q93077, (Human) P20671, (Human) Q9BTM1, (Human) P16104, (Human) P0C0S5, (Human) Q71UI9, (Mouse) Q8CGP5, (Mouse) C0HKE5, (Mouse) Q8CGP6, (Mouse) C0HKE6, (Mouse) Q8CGP7, (Mouse) Q6GSS7, (Mouse) Q64523, (Mouse) Q64522, (Mouse) C0HKE7, (Mouse) C0HKE8, (Mouse) C0HKE9, (Mouse) Q8BFU2, (Mouse) C0HKE1, (Mouse) C0HKE2, (Mouse) C0HKE3, (Mouse) C0HKE4, (Mouse) Q8R1M2, (Mouse) P27661, (Mouse) P0C0S6, (Mouse) Q3THW5

          View more View less

          Entrez Gene ID: (Human) 221613, (Human) 8969, (Human) 85235, (Human) 8329, (Human) 8331, (Human) 8330, (Human) 8332, (Human) 8336, (Human) 8337, (Human) 723790, (Human) 8338, (Human) 317772, (Human) 92815, (Human) 8335, (Human) 8334, (Human) 3013, (Human) 3012, (Human) 55766, (Human) 3014, (Human) 3015, (Human) 94239, (Mouse) 319163, (Mouse) 319173, (Mouse) 319167, (Mouse) 319168, (Mouse) 319191, (Mouse) 319169, (Mouse) 15267, (Mouse) 319192, (Mouse) 319176, (Mouse) 621893, (Mouse) 319170, (Mouse) 665433, (Mouse) 319171, (Mouse) 319162, (Mouse) 319172, (Mouse) 319164, (Mouse) 319165, (Mouse) 319166, (Mouse) 232440, (Mouse) 15270, (Mouse) 51788, (Mouse) 77605

          View more View less

          Function(s)
          DNA binding structural constituent of chromatin protein heterodimerization activity protein binding enzyme binding damaged DNA binding histone binding chromatin-protein adaptor activity RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II core promoter sequence-specific DNA binding chromatin DNA binding nucleosomal DNA binding binding, bridging
          Process(es)
          chromatin organization negative regulation of cell proliferation heterochromatin assembly protein localization to CENP-A containing chromatin positive regulation of transcription from RNA polymerase II promoter cellular response to estradiol stimulus DNA damage checkpoint double-strand break repair via homologous recombination DNA repair cellular response to DNA damage stimulus spermatogenesis protein K63-linked ubiquitination cellular response to gamma radiation positive regulation of double-strand break repair via homologous recombination protein localization to site of double-strand break double-strand break repair nucleosome assembly response to ionizing radiation positive regulation of DNA repair nucleosome disassembly UV-damage excision repair
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-tzrw4:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch