Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: STPSWCEEP AQANMDISTG HMILAYMEDH LRNRDRLAKE WQALCAYQAE PNTCATAQGE GNIKKNRHPD FLPYDHARIK LKVESSPSRS DYINASPIIE HDPRMPAYIA TQGPLSHTIA DFWQMVWESG CTVIVMLTPL VEDGVKQCDR YWPDEGASLY HVYEVNLVSE HIWCEDFLVR SFYLKNVQTQ ETRTLTQFHF LSWPAEGTPA STRPLLDFRR KVNKCYRGRS CPIIVHCSDG AGRTGTYILI DMVLNRMAKG VKEIDIAATL EHVRDQRPGL VRSKDQFEFA LTAVAEEVNA ILKALPQ (506-811 aa encoded by BC007713 )
IA-2 (insulinoma-associated protein 2, ICA512, PTPRN) is a member of the transmembrane protein tyrosine phosphatase (PTP) family located in secretory granules of neuroendocrine cells. The IA-2 protein is 979 amino acids in length and consists of a luminal domain, transmembrane domain, and cytoplasmic domain. Although a member of the PTP family, IA-2 lacks phosphatase activity with known substrates due to amino acid substitutions at critical sites in its PTP domain. Two paralog RPTPs, IA-2 and IA-2beta were identified as major autoantigens in type-1 diabetes mellitus. On granule exocytosis, the IA-2 cytoplasmic domain is cleaved and the resulting cytosolic fragment moves into the nucleus where it enhances the levels of phosphorylated STAT5 and STAT3, thereby inducing insulin gene transcription and granule biogenesis. IA-2 signaling enhances pancreatic beta-cell proliferation by regulating cyclins D through STATs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IA-2var; ICA 512; insulinoma-associated tyrosine-phosphatase-like protein; islet cell antigen 2; islet cell antigen 512; islet cell autoantigen 3; protein tyrosine phosphatase-like N; PTP IA-2; unnamed protein product
Gene Aliases: IA-2; IA-2/PTP; IA2; ICA512; R-PTP-N
UniProt ID: (Human) Q16849
Entrez Gene ID: (Human) 5798
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support