Applications | Tested Dilution | Publications |
---|---|---|
Flow Cytometry (Flow) |
10 µL per million cells | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
669222 |
Immunogen |
NS0 mouse myeloma cell line transfected with human ICOS and mouse myeloma cell line NS0-derived recombinant human ICOS cocktail Glu21-Phe141 |
Conjugate |
APC |
Excitation/Emission Profile |
View spectra |
Form |
Liquid |
Purification |
Protein A/G |
Storage buffer |
Buffered saline solution with BSA |
Contains |
0.09% sodium azide |
Storage conditions |
4° C, store in dark, DO NOT FREEZE! |
RRID |
AB_2576264 |
CD278 belongs to the CD28 and CTLA-4 cell-surface receptor family and is highly expressed in activated T-cells. It is essential both for efficient interaction between T and B-cells and for normal antibody responses to T-cell dependent antigens. It enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: Activation-inducible lymphocyte immunomediatory molecule; CD278; inducible costimulator; inducible T-cell co-stimulator; Inducible T-cell costimulator
Gene Aliases: AILIM; CD278; CVID1; ICOS
UniProt ID: (Human) Q9Y6W8
Entrez Gene ID: (Human) 29851
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support