Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:2,000-1:8,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
3D8B11 |
Immunogen |
IFIH1 Fusion Protein Ag17448 (1-310 aa encoded by BC111750) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein G |
Storage buffer |
PBS, pH 7.3, with 50% glycerol |
Contains |
0.1% sodium azide |
Storage conditions |
-20°C |
Immunogen sequence: MSNGYSTDE NFRYLISCFR ARVKMYIQVE PVLDYLTFLP AEVKEQIQRT VATSGNMQAV ELLLSTLEKG VWHLGWTREF VEALRRTGSP LAARYMNPEL TDLPSPSFEN AHDEYLQLLN LLQPTLVDKL LVRDVLDKCM EEELLTIEDR NRIAAAENNG NESGVRELLK RIVQKENWFS AFLNVLRQTG NNELVQELTG SDCSESNAEI ENLSQVDGPQ VEEQLLSTTV QPNLEKEVWG MENNSSESSF ADSSVVSESD TSLAEGSVSC LDESLGHNSN MGSDSGTMGS DSDEENVAAR ASPEPELQLR P (1-310 aa encoded by BC111750)
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp and a protein kinase C-activating compound, mezerein. Irreversible reprogramming of melanomas can be achieved by treatment with both these agents; treatment with either agent alone only achieves reversible differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CADM-140 autoantigen; Clinically amyopathic dermatomyositis autoantigen 140 kDa; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; Helicard; Helicase with 2 CARD domains; interferon induced, with helicase C domain 1; Interferon-induced helicase C domain-containing protein 1; Interferon-induced with helicase C domain protein 1; MDA-5; melanoma differentiation associated protein-5; melanoma differentiation-associated gene 5; Melanoma differentiation-associated protein 5; MGC133047; Murabutide down-regulated protein; RIG-I-like receptor 2; RLR-2; RNA helicase-DEAD box protein 116
Gene Aliases: AGS7; Hlcd; IDDM19; IFIH1; MDA-5; MDA5; RH116; RLR-2; SGMRT1
UniProt ID: (Human) Q9BYX4
Entrez Gene ID: (Human) 64135
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support