Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Flow Cytometry (Flow) |
- | View 1 publication 1 publication |
ELISA (ELISA) |
1-4 µg/mL | View 17 publications 17 publications |
Functional Assay (FN) |
Assay-Dependent | View 3 publications 3 publications |
Control (Ctrl) |
Assay-Dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Mouse |
Published species |
Human, Mouse |
Host/Isotype |
Rat / IgG1, kappa |
Recommended Isotype Control |
Rat IgG1 kappa Isotype Control (eBRG1), Functional Grade, eBioscience™ |
Class |
Monoclonal |
Type |
Antibody |
Clone |
AN-18 |
Conjugate |
Functional Grade
|
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.2 |
Contains |
no preservative |
Storage conditions |
4° C |
RRID |
AB_469246 |
Description: The AN-18 antibody reacts with mouse interferon-gamma (IFN gamma), a 15-20 kDa cytokine secreted by Th1 cells.
Applications Reported: The AN-18 antibody has been reported for use in ELISA and ELISPOT assay. For neutralization of IFN gamma activity, the alternate clones R4-6A2 or XMG1.2 are recommended.
Applications Tested: The AN-18 antibody has been tested as the capture antibody in IFN gamma ELISPOT assay, in combination with the biotinylated R4-6A2 antibody for detection. A concentration of 1-4 µg/mL is recommended. For neutralization of IFN gamma activity, the alternate clones R4-6A2 or XMG1.2 are recommended.
Storage and handling: Use in a sterile environment.
Filtration: 0.2 µm post-manufacturing filtered.
Purity: Greater than 90%, as determined by SDS-PAGE.
Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.
Aggregation: Less than 10%, as determined by HPLC.
IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gamma interferon; H-IFN-g; IFN γ; IFN-gamma; IFN-y; IFNγ; Interferon gamma; Interferon y; Interferon γ; Interferonγ; M-IFN-g; R-IFN-g
Gene Aliases: Ifg; IFN-g; Ifng
UniProt ID: (Mouse) P01580
Entrez Gene ID: (Mouse) 15978
Molecular Function:
cytokine
interferon superfamily
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support