Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:10-1:200 | View 3 publications 3 publications |
Immunohistochemistry (Paraffin) (IHC (P)) |
- | View 1 publication 1 publication |
Immunocytochemistry (ICC/IF) |
1:10-1:100 | View 1 publication 1 publication |
Immunoprecipitation (IP) |
2 µg/mL | View 2 publications 2 publications |
ChIP assay (ChIP) |
- | View 1 publication 1 publication |
Inhibition Assays (IA) |
1.5 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Published species |
Human |
Host/Isotype |
Mouse / IgG2a, kappa |
Class |
Monoclonal |
Type |
Antibody |
Clone |
24-60 |
Immunogen |
IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.2 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS, pH 7.4, with 0.2% BSA |
Contains |
0.09% sodium azide |
Storage conditions |
4° C |
RRID |
AB_10986755 |
MA5-13817 targets IGF-1 Receptor alpha in IA, ICC/IF, WB and IP applications and shows reactivity with Human and Mouse samples.
The MA5-13817 immunogen is iGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA.
IGF1R (Insulin-like Growth Factor1 Receptor) is a receptor tyrosine kinase that mediates actions of insulin-like growth factor 1 (IFG1). It binds IGF1 with high affinity and IGF2/insulin with lower affinity. Activated IGF1R is involved in cell growth and survival control. IGF1R is crucial for tumor transformation and survial of malignant cells. Ligand binding activates the receptor kinase and leads to receptor autophosphorylation and tyrosines phosphorylation of multiple substrates that function as signaling adapter proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD221; IGF-I receptor; Insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Aliases: A330103N21Rik; CD221; D930020L01; hyft; IGF-1R; IGF1R; IGFIR; IGFR; JTK13
UniProt ID: (Human) P08069, (Mouse) Q60751
Entrez Gene ID: (Human) 3480, (Mouse) 16001
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support