Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SPGQGTQSE NSCTHFPGNL PNMLRDLRDA FSRVKTFFQM KDQLDNLLLK ESLLEDFKGY LGCQALSEMI QFYLEEVMPQ AENQDPDIKA HVNSLGENLK TLRLRLRRCH RFLPCENKSK AVEQVKNAFN KLQEKGIYKA MSEFDIFINY IEAYMTMKIR N (19-178 aa encoded by BC104252)
Interleukin 10 (IL-10, CSIF) is an anti-inflammatory cytokine mainly produced by macrophages and Th2 cells. IL-10 is an 18.6 kDa protein of 160 amino acid residues that shares over 80% sequence homology with the Epstein-Barr Virus protein (BCRFI). The reported biological activities of IL-10, which maybe interrelated, include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hyper-sensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production. IL-10 functions by inhibiting pro-inflammatory cytokines made by macrophages and regulatory T cells including IFN-gamma, TNF-alpha, IL-2, and IL-3, IL-4, and GM-CSF. IL-10 is also known to suppress antigen presentation on antigen presenting cells, enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. While human IL-10 can act on murine cells, murine IL-10 cannot act on human cells. Binds to a retinoid X receptor (RXR) responsive element from the cellular retinol-binding protein II promoter (CRBPII-RXRE). IL-10 inhibits the 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element. Knockout studies in mice suggest the function of this cytokine as an essential immunoregulator in the intestinal tract.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CSIF; Cytokine synthesis inhibitory factor; H-IL-10; il 10; IL-10; ILN; Interleukin; Interleukin-10; Interleukin10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor
Gene Aliases: IL10
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support