Invitrogen
Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for this product is human tonsil.
For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
Rat anti Human Interleukin-10 antibody, clone JES3-12G8 recognizes human Interleukin-10 (IL-10).
Interleukin 10 (IL-10, CSIF) is an anti-inflammatory cytokine mainly produced by macrophages and Th2 cells. IL-10 is an 18.6 kDa protein of 160 amino acid residues that shares over 80% sequence homology with the Epstein-Barr Virus protein (BCRFI). The reported biological activities of IL-10, which maybe interrelated, include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hyper-sensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production. IL-10 functions by inhibiting pro-inflammatory cytokines made by macrophages and regulatory T cells including IFN-gamma, TNF-alpha, IL-2, and IL-3, IL-4, and GM-CSF. IL-10 is also known to suppress antigen presentation on antigen presenting cells, enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. While human IL-10 can act on murine cells, murine IL-10 cannot act on human cells. Binds to a retinoid X receptor (RXR) responsive element from the cellular retinol-binding protein II promoter (CRBPII-RXRE). IL-10 inhibits the 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element. Knockout studies in mice suggest the function of this cytokine as an essential immunoregulator in the intestinal tract.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CSIF; Cytokine synthesis inhibitory factor; H-IL-10; il 10; IL-10; ILN; Interleukin; Interleukin-10; Interleukin10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor
Gene Aliases: CSIF; GVHDS; IL-10; IL10; IL10A; TGIF
UniProt ID: (Human) P22301
Entrez Gene ID: (Human) 3586
Molecular Function:
cytokine
interleukin superfamily
signaling molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support