Product References
RORγt phosphorylation protects against T cell-mediated inflammation.
Cell reports
Ma S,Patel SA,Abe Y,Chen N,Patel PR,Cho BS,Abbasi N,Zeng S,Schnabl B,Chang JT,Huang WJM
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Tue Mar 15 00:00:00 EDT 2022
Milk-derived extracellular vesicles alleviate ulcerative colitis by regulating the gut immunity and reshaping the gut microbiota.
Theranostics
Tong L,Hao H,Zhang Z,Lv Y,Liang X,Liu Q,Liu T,Gong P,Zhang L,Cao F,Pastorin G,Lee CN,Chen X,Wang JW,Yi H
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Fri Jan 28 00:00:00 EST 2022
Characterisation of Macrophage Polarisation in Mice Infected with Ninoa Strain of Trypanosoma cruzi.
Pathogens (Basel, Switzerland)
Medina-Buelvas DM,Rodríguez-Sosa M,Vega L
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Sat Nov 06 00:00:00 EDT 2021
Gut epithelial IL-27 confers intestinal immunity through the induction of intraepithelial lymphocytes.
The Journal of experimental medicine
Lin CH,Chen MC,Lin LL,Christian DA,Min B,Hunter CA,Lu LF
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Mon Nov 01 00:00:00 EDT 2021
Irradiated whole-cell vaccine suppresses hepatocellular carcinoma growth in mice via Th9 cells.
Oncology letters
Chen J,Ding Y,Huang F,Lan R,Wang Z,Huang W,Chen R,Wu B,Fu L,Yang Y,Liu J,Hong J,Zhang W,Zhang L
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Sat May 01 00:00:00 EDT 2021
TGF-β1 secreted by Tregs in lymph nodes promotes breast cancer malignancy via up-regulation of IL-17RB.
EMBO molecular medicine
Huang SC,Wei PC,Hwang-Verslues WW,Kuo WH,Jeng YM,Hu CM,Shew JY,Huang CS,Chang KJ,Lee EY,Lee WH
16-7101 was used in Flow cytometry/Cell sorting to investigate the role that regulatory T cells (in tumour-draining lymph nodes) play in modulating the malignancy of breast cancer cells for distant metastasis.
Fri Dec 01 00:00:00 EST 2017
NOX2 deficiency alters macrophage phenotype through an IL-10/STAT3 dependent mechanism: implications for traumatic brain injury.
Journal of neuroinflammation
Barrett JP,Henry RJ,Villapol S,Stoica BA,Kumar A,Burns MP,Faden AI,Loane DJ
16-7101 was used in Neutralization experiments to investigate the signalling pathways that regulate NOX2-dependent macrophage/microglial phenotype switching in the TBI brain.
Fri Mar 24 00:00:00 EDT 2017
Excessive expression of miR-27 impairs Treg-mediated immunological tolerance.
The Journal of clinical investigation
Cruz LO,Hashemifar SS,Wu CJ,Cho S,Nguyen DT,Lin LL,Khan AA,Lu LF
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
Heterologous Immunity between Adenoviruses and Hepatitis C Virus: A New Paradigm in HCV Immunity and Vaccines.
PloS one
Singh S,Vedi S,Samrat SK,Li W,Kumar R,Agrawal B
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Tue Jul 19 00:00:00 EDT 2016
NFATc1 supports imiquimod-induced skin inflammation by suppressing IL-10 synthesis in B cells.
Nature communications
Alrefai H,Muhammad K,Rudolf R,Pham DA,Klein-Hessling S,Patra AK,Avots A,Bukur V,Sahin U,Tenzer S,Goebeler M,Kerstan A,Serfling E
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Wed May 25 00:00:00 EDT 2016
Maternal-Derived Hepatitis B Virus e Antigen Alters Macrophage Function in Offspring to Drive Viral Persistence after Vertical Transmission.
Immunity
Tian Y,Kuo CF,Akbari O,Ou JH
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Tue May 17 00:00:00 EDT 2016
Towards programming immune tolerance through geometric manipulation of phosphatidylserine.
Biomaterials
Roberts RA,Eitas TK,Byrne JD,Johnson BM,Short PJ,McKinnon KP,Reisdorf S,Luft JC,DeSimone JM,Ting JP
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2015
Tristetraprolin Limits Inflammatory Cytokine Production in Tumor-Associated Macrophages in an mRNA Decay-Independent Manner.
Cancer research
Kratochvill F,Gratz N,Qualls JE,Van De Velde LA,Chi H,Kovarik P,Murray PJ
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2015
Mouse bone marrow-derived mesenchymal stem cells inhibit leukemia/lymphoma cell proliferation in vitro and in a mouse model of allogeneic bone marrow transplant.
International journal of molecular medicine
Song N,Gao L,Qiu H,Huang C,Cheng H,Zhou H,Lv S,Chen L,Wang J
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in ELISA
Wed Jul 01 00:00:00 EDT 2015
Mouse bone marrow-derived mesenchymal stem cells inhibit leukemia/lymphoma cell proliferation in vitro and in a mouse model of allogeneic bone marrow transplant.
International journal of molecular medicine
Song N,Gao L,Qiu H,Huang C,Cheng H,Zhou H,Lv S,Chen L,Wang J
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in ELISA
Wed Jul 01 00:00:00 EDT 2015
Canonical wnt signaling in dendritic cells regulates Th1/Th17 responses and suppresses autoimmune neuroinflammation.
Journal of immunology (Baltimore, Md. : 1950)
Suryawanshi A,Manoharan I,Hong Y,Swafford D,Majumdar T,Taketo MM,Manicassamy B,Koni PA,Thangaraju M,Sun Z,Mellor AL,Munn DH,Manicassamy S
16-7101 was used in Flow cytometry/Cell sorting to study the factors in the tissue microenvironment and signalling networks critical for programming dendritic cells to control chronic inflammation and promote tolerance, showing that Th1/Th17 responses are regulated by canonical Wnt signalling in dendritic cells and suppresses autoimmune neuroinflammation.
Wed Apr 01 00:00:00 EDT 2015
Nizatidine, a small molecular compound, enhances killed H5N1 vaccine cell-mediated responses and protects mice from lethal viral challenge.
Human vaccines & immunotherapeutics
Wang S,Wu B,Xue J,Wang M,Chen R,Wang B
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Mon Jan 05 00:00:00 EST 2015
Gr1(int)CD11b+ myeloid-derived suppressor cells in Mycobacterium tuberculosis infection.
PloS one
Obregón-Henao A,Henao-Tamayo M,Orme IM,Ordway DJ
16-7101 was used in Flow cytometry/Cell sorting to compare the bacterial burden, lung pathology and Gr1(int)CD11b(+) myeloid-derived suppressor cell immune responses in M. tuberculosis infected mice.
Fri Jun 20 00:00:00 EDT 2014
CD3 antibody and IL-2 complex combination therapy inhibits atherosclerosis by augmenting a regulatory immune response.
Journal of the American Heart Association
Kasahara K,Sasaki N,Yamashita T,Kita T,Yodoi K,Sasaki Y,Takeda M,Hirata K
16-7101 was used in Flow cytometry/Cell sorting to investigate the inhibition of atherosclerosis by a CD3 antibody and IL-2 complex combination.
Tue Apr 22 00:00:00 EDT 2014
BCG vaccine mediated reduction in the MHC-II expression of macrophages and dendritic cells is reversed by activation of Toll-like receptors 7 and 9.
Cellular immunology
Bakhru P,Sirisaengtaksin N,Soudani E,Mukherjee S,Khan A,Jagannath C
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2014
Role of arginase 1 from myeloid cells in th2-dominated lung inflammation.
PloS one
Barron L,Smith AM,El Kasmi KC,Qualls JE,Huang X,Cheever A,Borthwick LA,Wilson MS,Murray PJ,Wynn TA
16-7101 was used in Flow cytometry/Cell sorting to investigate the contribution of arginase 1 to asthma pathogenesis.
Mon Dec 09 00:00:00 EST 2013
Comparative analysis of cancer vaccine settings for the selection of an effective protocol in mice.
Journal of translational medicine
Kalli F,Machiorlatti R,Battaglia F,Parodi A,Conteduca G,Ferrera F,Proietti M,Tardito S,Sanguineti M,Millo E,Fenoglio D,De Palma R,Inghirami G,Filaci G
16-7101 was used in a Functional assay to compare different gp100 vaccination strategies to identify the best strategy to provide a 100% protection against experimental melanoma in a reproducible manner.
Sun May 12 00:00:00 EDT 2013
Genetic deletion of chemokine receptor Ccr7 exacerbates atherogenesis in ApoE-deficient mice.
Cardiovascular research
Wan W,Lionakis MS,Liu Q,Roffê E,Murphy PM
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
CX(3)CR1(+) macrophages support IL-22 production by innate lymphoid cells during infection with Citrobacter rodentium.
Mucosal immunology
Manta C,Heupel E,Radulovic K,Rossini V,Garbi N,Riedel CU,Niess JH
Published figure using IL-10 monoclonal antibody (Product # 16-7101-81) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2013
The E3 ubiquitin ligase adaptor Ndfip1 regulates Th17 differentiation by limiting the production of proinflammatory cytokines.
Journal of immunology (Baltimore, Md. : 1950)
Ramon HE,Beal AM,Liu Y,Worthen GS,Oliver PM
16-7101 was used in Flow cytometry/Cell sorting to investigate the role of the E3 ubiquitin ligase adaptor Ndfip1 on Th17 differentiation in mice.
Sun Apr 15 00:00:00 EDT 2012
Novel CCL21-vault nanocapsule intratumoral delivery inhibits lung cancer growth.
PloS one
Kar UK,Srivastava MK,Andersson A,Baratelli F,Huang M,Kickhoefer VA,Dubinett SM,Rome LH,Sharma S
16-7101 was used in Flow cytometry/Cell sorting to demonstrate that the vault nanocapsule can efficiently deliver CCL21 to recruit antitumour effectors, sustain antitumour activity and inhibit tumour growth.
Tue May 03 00:00:00 EDT 2011
Separation of graft-versus-host disease from graft-versus-leukemia responses by targeting CC-chemokine receptor 7 on donor T cells.
Blood
Coghill JM,Carlson MJ,Panoskaltsis-Mortari A,West ML,Burgents JE,Blazar BR,Serody JS
16-7101 was used in Flow cytometry/Cell sorting to investigate the role of MHC disparity between donor and recipient, in the attenuated graft-versus-host disease responses of murine T cells lacking CCR7.
Thu Jun 10 00:00:00 EDT 2010
Synthetic cationic peptide IDR-1002 provides protection against bacterial infections through chemokine induction and enhanced leukocyte recruitment.
Journal of immunology (Baltimore, Md. : 1950)
Nijnik A,Madera L,Ma S,Waldbrook M,Elliott MR,Easton DM,Mayer ML,Mullaly SC,Kindrachuk J,Jenssen H,Hancock RE
16-7101 was used in an ELISA assay to study the importance of the chemokine induction activity of host defence peptides in protection against bacterial infections.
Mon Mar 01 00:00:00 EST 2010
Role of plasmacytoid dendritic cells for aberrant class II expression in exocrine glands from estrogen-deficient mice of healthy background.
The American journal of pathology
Arakaki R,Nagaoka A,Ishimaru N,Yamada A,Yoshida S,Hayashi Y
16-7101 was used in an ELISA assay to indicate that estrogen deficiency initiates class II transactivator type IV mRNA expression in exocrine glands via epithelial and plasmacytoid DC interactions.
Fri May 01 00:00:00 EDT 2009