Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL-10 Antibodies

          Invitrogen

          IL-10 Monoclonal Antibody (JES5-16E3), PerCP-Cyanine5.5, eBioscience™

          27 Published Figures
          26 References
          View all (112) IL-10 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IL-10 Monoclonal Antibody (JES5-16E3), PerCP-Cyanine5.5, eBioscience™

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (27)
          IL-10 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          IL-10 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 28

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-10 Antibody (45-7101-82) in Flow

          Intracellular staining of Mouse Cytokine Positive Control Cells (Product # 00-4500-51) with Anti-Mouse CD4 PE (Product # 12-0042-82) and 0.25 µg of Rat IgG2b K Isotype Control PerCP-Cyanine5-5 (Product # 45-4031-80) (left) or 0.25 µg of Anti-Mouse IL-10 PerCP-Cyanine5-5 (right). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)
          IL-10 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          45-7101-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 25 publications 25 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), PerCP-Cyanine5.5, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          JES5-16E3

          Conjugate

          PerCP-Cyanine5.5 PerCP-Cyanine5.5 PerCP-Cyanine5.5
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 700 
          • APC 
          • Biotin 
          • FITC
          • Functional Grade
          • PE 
          • Request custom conjugation

          Excitation/Emission Max

          489/679 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_996677

          Product Specific Information

          Description: The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). Mouse IL-10 is an ~18 kDa factor also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells, macrophages, and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response.

          Applications Reported: This JES5-16E3 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

          Applications Tested: This JES5-16E3 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: cytokine synthesis inhibitory factor; H-IL-10; il 10; ILN; interferon 2a; Interleukin; interleukin 10 precursor; Interleukin10; MGC126450; MGC126451; RP11-262N9.1; unnamed protein product

          View more View less

          Gene Aliases: CSIF; If2a; Il-10

          View more View less

          UniProt ID: (Mouse) P18893

          View more View less

          Entrez Gene ID: (Mouse) 16153

          View more View less

          Function(s)
          cytokine activity interleukin-10 receptor binding protein dimerization activity
          Process(es)
          negative regulation of cytokine production positive regulation of cytokine production positive regulation of endothelial cell proliferation response to molecule of bacterial origin positive regulation of immunoglobulin production negative regulation of cytokine production involved in immune response negative regulation of chronic inflammatory response to antigenic stimulus positive regulation of B cell apoptotic process immune response negative regulation of cell proliferation response to xenobiotic stimulus regulation of gene expression negative regulation of autophagy response to activity response to inactivity negative regulation of myeloid dendritic cell activation negative regulation of B cell proliferation response to lipopolysaccharide negative regulation of interferon-gamma production negative regulation of interleukin-12 production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production response to insulin negative regulation of heterotypic cell-cell adhesion response to carbon monoxide cellular response to hepatocyte growth factor stimulus defense response to bacterium defense response to protozoan positive regulation of macrophage activation negative regulation of apoptotic process negative regulation of neuron apoptotic process negative regulation of nitric oxide biosynthetic process T-helper 1 cell differentiation negative regulation of MHC class II biosynthetic process positive regulation of MHC class II biosynthetic process positive regulation of cell cycle positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JAK-STAT cascade negative regulation of inflammatory response regulation of synapse organization regulation of B cell activation negative regulation of membrane protein ectodomain proteolysis response to glucocorticoid negative regulation of cytokine activity branching involved in labyrinthine layer morphogenesis cellular response to lipopolysaccharide cellular response to estradiol stimulus liver regeneration interleukin-10-mediated signaling pathway positive regulation of plasma cell differentiation positive regulation of pri-miRNA transcription from RNA polymerase II promoter regulation of response to wounding negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway positive regulation of sprouting angiogenesis negative regulation of vascular smooth muscle cell proliferation negative regulation of endothelial cell apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-p95z8:80/100.66.74.151:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline