Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MM120 targets IL-12 in ELISA, and WB applications and shows reactivity with mouse samples.
The MM120 immunogen is recombinant mouse IL-12.
MM120 detects IL-12 which has a predicted molecular weight of approximately 22 kDa.
The MM120 mouse IL12 antibody (clone 9A5) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody MM121B (biotinylated conjugate of clone C17-8). Typical dilutions for sandwich ELISA include 1 µg/mL for coating and 0.125 - 0.25 µg/mL for detection.
Antibody MM120 (clone 9A5) and biotinylated antibody MM121B (clone C17-8) have successfully been used in combination with recombinant mouse IL12p70 protein SMIL12 in ELISA applications.
This antibody is produced by injecting Rat IgG secreting hybridoma cells into the peritoneum of mice. The resulting ascites is collected from the mice and the antibody is purified.
This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 20C2; CLMF p35; CLMF p40; cytotoxic lymphocyte maturation factor 35 kDa subunit; cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12p70; IL-12 p35 subunit; IL-12 p40 subunit; IL-12 p75; IL-12 subunit p35; IL-12 subunit p40; IL-23 subunit p40; IL12A/B; IL12AB; ILN; Interleukin; interleukin 12a p35 subunit; interleukin-12 p35 subunit; interleukin-12 p40 subunit; Interleukin-12 p70; MGC151228; MGC151232; unnamed protein product
Gene Aliases: Il-12a; Il-12b; IL-12p35; Il-12p40; Il12p40; Ll12a; p35; p40
UniProt ID: (Mouse) P43431, (Mouse) P43432
Entrez Gene ID: (Mouse) 16159, (Mouse) 16160
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support