Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • IL-12 p70 Antibodies

          Invitrogen

          IL-12 p70 Monoclonal Antibody (A25C4B6), Biotin

          1 Published Figure
          3 References
          View all (26) IL-12 p70 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite IL-12 p70 Monoclonal Antibody (A25C4B6), Biotin

          • Published Figures (1)
          IL-12 p70 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          IL-12 p70 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-12 p70 Antibody (AHC7129) in Flow

          EBioMedicine 2019 - Fig. 1 Increased plasma levels of IL-12 and IL-6 correlate with increased non-classical CD16 bright CD14 neg monocytes as well as with increased activation of classical CD16 neg CD14 bright monocytes in patients with MDD and suicide behaviour. Plasma levels of (a) IL-12p70 and (b) IL-6 in patients with MDD (n=33) and healthy controls (n=20) as determined by ELISA. Significant differences between patients wi... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-12 p70 Antibody in Flow Cytometry (Flow)

          Product Details

          AHC7129

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-dependent
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          A25C4B6

          Immunogen

          Recombinant human IL-12.

          Conjugate

          Biotin Biotin Biotin

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 5mg/mL BSA

          Contains

          0.5mg/mL sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2536290

          Product Specific Information

          This product is recommended as a detection antibody in Sandwich ELISA applications.

          This clone can also be referred to as clone number A25C 4B6 1B3.

          Target Information

          IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 20C2; CLMF p35; CLMF p40; cytotoxic lymphocyte maturation factor 1, p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12p70; IL-12 p75; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL-12A; IL-12B; IL12, subunit p40; IL12A/B; IL12AB; IL35 subunit; ILN; Interleukin; interleukin 12, p35; interleukin 12, p40; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 alpha chain; interleukin-12 beta chain; Interleukin-12 p70; Interleukin-12 subunit alpha; Interleukin-12 subunit beta; MGC151228; MGC151232; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF1; NKSF2

          View more View less

          Gene Aliases: CLMF; CLMF2; IL-12A; IL-12B; IL12A; IL12B; IMD28; IMD29; NFSK; NKSF; NKSF1; NKSF2; P35

          View more View less

          UniProt ID: (Human) P29459, (Human) P29460

          View more View less

          Entrez Gene ID: (Human) 3592, (Human) 3593

          View more View less

          Function(s)
          cytokine activity interleukin-12 receptor binding protein binding growth factor activity interleukin-12 beta subunit binding interleukin-27 binding protein heterodimerization activity cytokine receptor activity interleukin-12 alpha subunit binding identical protein binding protein homodimerization activity interleukin-23 receptor binding transmembrane signal receptor
          Process(es)
          positive regulation of T cell mediated cytotoxicity positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target immune response cell cycle arrest response to virus response to UV-B cell migration response to lipopolysaccharide negative regulation of interleukin-17 production positive regulation of interferon-gamma production positive regulation of natural killer cell activation positive regulation of mononuclear cell proliferation positive regulation of smooth muscle cell apoptotic process positive regulation of T cell proliferation positive regulation of tyrosine phosphorylation of Stat4 protein defense response to protozoan positive regulation of T cell differentiation positive regulation of cell adhesion positive regulation of natural killer cell mediated cytotoxicity negative regulation of smooth muscle cell proliferation positive regulation of lymphocyte proliferation defense response to Gram-positive bacterium positive regulation of NK T cell activation cellular response to lipopolysaccharide extrinsic apoptotic signaling pathway cellular response to virus positive regulation of dendritic cell chemotaxis positive regulation of defense response to virus by host natural killer cell activation involved in immune response positive regulation of T-helper 1 type immune response negative regulation of inflammatory response to antigenic stimulus positive regulation of activation of JAK2 kinase activity cytokine-mediated signaling pathway sensory perception of pain sexual reproduction natural killer cell activation negative regulation of interleukin-10 production positive regulation of granulocyte macrophage colony-stimulating factor production positive regulation of interleukin-10 production positive regulation of interleukin-12 production positive regulation of interleukin-17 production positive regulation of tumor necrosis factor production positive regulation of natural killer cell proliferation positive regulation of tissue remodeling regulation of cytokine biosynthetic process T-helper 1 type immune response T-helper cell differentiation interferon-gamma biosynthetic process positive regulation of activated T cell proliferation positive regulation of NF-kappaB import into nucleus regulation of tyrosine phosphorylation of Stat1 protein positive regulation of tyrosine phosphorylation of Stat3 protein positive regulation of tyrosine phosphorylation of Stat5 protein positive regulation of memory T cell differentiation negative regulation of growth of symbiont in host positive regulation of interferon-gamma biosynthetic process positive regulation of osteoclast differentiation positive regulation of inflammatory response defense response to Gram-negative bacterium positive regulation of NK T cell proliferation defense response to virus cellular response to interferon-gamma positive regulation of T-helper 17 type immune response positive regulation of T-helper 17 cell lineage commitment
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-57b588f4f5-kdjgd:80/100.66.79.206:80.
          git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.39.0-Offline