Product References
Vitamin D3 mitigates autoimmune inflammation caused by activation of myeloid dendritic cells in SLE.
Experimental dermatology
Li M,Luo L,Lin C,Ni B,Zou L,Song Z,Hao F,Wu Y,Luo N
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Mon Jan 01 00:00:00 UTC 2024
FABP4 facilitates inflammasome activation to induce the Treg/Th17 imbalance in preeclampsia via forming a positive feedback with IL-17A.
Molecular therapy. Nucleic acids
Chang GP,Yang XL,Liu W,Lin S,Yang SL,Zhao MY
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Neutralization
Fri Jun 04 00:00:00 UTC 2021
Allogeneic ADSCs Induce the Production of Alloreactive Memory-CD8 T Cells through HLA-ABC Antigens.
Cells
Chang SH,Kim HJ,Park CG
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Neutralization
Mon May 18 00:00:00 UTC 2020
Th2 Biased Immunity With Altered B Cell Profiles in Circulation of Patients With Sporotrichosis Caused by Sporothrix globosa.
Frontiers in immunology
Zu J,Yao L,Song Y,Cui Y,Guan M,Chen R,Zhen Y,Li S
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Mon May 31 00:00:00 UTC 2021
Allogeneic ADSCs induce CD8 T cell-mediated cytotoxicity and faster cell death after exposure to xenogeneic serum or proinflammatory cytokines.
Experimental & molecular medicine
Chang SH,Park CG
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Mon Mar 11 00:00:00 UTC 2019
Allogeneic ADSCs induce CD8 T cell-mediated cytotoxicity and faster cell death after exposure to xenogeneic serum or proinflammatory cytokines.
Experimental & molecular medicine
Chang SH,Park CG
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Mon Mar 11 00:00:00 UTC 2019
Tim-3 regulates inflammatory cytokine expression and Th17 cell response induced by monocytes from patients with chronic hepatitis B.
Scandinavian journal of immunology
Wang J,Li C,Fu J,Wang X,Feng X,Pan X
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Wed May 01 00:00:00 UTC 2019
P2X7R mutation disrupts the NLRP3-mediated Th program and predicts poor cardiac allograft outcomes.
The Journal of clinical investigation
D'Addio F,Vergani A,Potena L,Maestroni A,Usuelli V,Ben Nasr M,Bassi R,Tezza S,Dellepiane S,El Essawy B,Iascone M,Iacovoni A,Borgese L,Liu K,Visner G,Dhe-Paganon S,Corradi D,Abdi R,Starling RC,Folli F,Zuccotti GV,Sayegh MH,Heeger PS,Chandraker A,Grigioni F,Fiorina P
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Wed Aug 01 00:00:00 UTC 2018
Clonally expanded γδ T cells protect against Staphylococcus aureus skin reinfection.
The Journal of clinical investigation
Dillen CA,Pinsker BL,Marusina AI,Merleev AA,Farber ON,Liu H,Archer NK,Lee DB,Wang Y,Ortines RV,Lee SK,Marchitto MC,Cai SS,Ashbaugh AG,May LS,Holland SM,Freeman AF,Miller LG,Yeaman MR,Simon SI,Milner JD,Maverakis E,Miller LS
14-7178 was used in Flow cytometry/Cell sorting to demonstrate that γδ T cells that are clonally explanded represent a mechanism for long-lasting immunity against recurrent S. aureus skin infections.
Thu Mar 01 00:00:00 UTC 2018
The Effects of High Mobility Group Box-1 Protein on Peripheral Treg/Th17 Balance in Patients with Atherosclerosis.
Acta Cardiologica Sinica
Ding JW,Zhou T,Zheng XX,Wang XA,Tong XH,Luo CY,Zhang ZQ,Yu B
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Sat Sep 01 00:00:00 UTC 2018
Application of a whole blood mycobacterial growth inhibition assay to study immunity against Mycobacterium tuberculosis in a high tuberculosis burden population.
PloS one
Baguma R,Penn-Nicholson A,Smit E,Erasmus M,Day J,Makhethe L,de Kock M,Hughes EJ,van Rooyen M,Pienaar B,Stone L,Hanekom W,Brennan MJ,Wallis RS,Hatherill M,Scriba TJ
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Wed Oct 18 00:00:00 UTC 2017
Balancing Trained Immunity with Persistent Immune Activation and the Risk of Simian Immunodeficiency Virus Infection in Infant Macaques Vaccinated with Attenuated Mycobacterium tuberculosis or Mycobacterium bovis BCG Vaccine.
Clinical and vaccine immunology : CVI
Jensen K,Dela Pena-Ponce MG,Piatak M,Shoemaker R,Oswald K,Jacobs WR,Fennelly G,Lucero C,Mollan KR,Hudgens MG,Amedee A,Kozlowski PA,Estes JD,Lifson JD,Van Rompay KK,Larsen M,De Paris K
14-7178 was used in Flow cytometry/Cell sorting to develop a pediatric combination vaccine to protect the vulnerable infant population against human immunodeficiency virus type 1 (HIV-1) and tuberculosis (TB) infections.
Sun Jan 01 00:00:00 UTC 2017
Transcriptional Profiling of Th2 Cells Identifies Pathogenic Features Associated with Asthma.
Journal of immunology (Baltimore, Md. : 1950)
Seumois G,Zapardiel-Gonzalo J,White B,Singh D,Schulten V,Dillon M,Hinz D,Broide DH,Sette A,Peters B,Vijayanand P
14-7178 was used in an ELISA assay to identify several molecules that are likely to confer pathogenic features to Th2 cells that are either unique or common to both asthma and rhinitis.
Fri Jul 15 00:00:00 UTC 2016
Preclinical Testing of Antihuman CD28 Fab' Antibody in a Novel Nonhuman Primate Small Animal Rodent Model of Xenogenic Graft-Versus-Host Disease.
Transplantation
Hippen KL,Watkins B,Tkachev V,Lemire AM,Lehnen C,Riddle MJ,Singh K,Panoskaltsis-Mortari A,Vanhove B,Tolar J,Kean LS,Blazar BR
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Thu Dec 01 00:00:00 UTC 2016
Altered expression of miR-92a correlates with Th17 cell frequency in patients with primary biliary cirrhosis.
International journal of molecular medicine
Liang DY,Hou YQ,Luo LJ,Ao L
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Fri Jul 01 00:00:00 UTC 2016
Effects of B Cell Depletion on Early Mycobacterium tuberculosis Infection in Cynomolgus Macaques.
Infection and immunity
Phuah J,Wong EA,Gideon HP,Maiello P,Coleman MT,Hendricks MR,Ruden R,Cirrincione LR,Chan J,Lin PL,Flynn JL
14-7178 was used in Flow cytometry/Cell sorting to examine the contribution of B cells and humoural immunity to the control of tuberculosis in non-human primates during the acute phase of infection.
Sun May 01 00:00:00 UTC 2016
Downregulation of RUNX3 moderates the frequency of Th17 and Th22 cells in patients with psoriasis.
Molecular medicine reports
Fu D,Song X,Hu H,Sun M,Li Z,Tian Z
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Wed Jun 01 00:00:00 UTC 2016
A molecular signature of preclinical rheumatoid arthritis triggered by dysregulated PTPN22.
JCI insight
Chang HH,Liu GY,Dwivedi N,Sun B,Okamoto Y,Kinslow JD,Deane KD,Demoruelle MK,Norris JM,Thompson PR,Sparks JA,Rao DA,Karlson EW,Hung HC,Holers VM,Ho IC
14-7178 was used in an ELISA assay to depict a molecular signature of preclinical rheumatoid arthritis that is triggered by impaired induction of PTPN22.
Thu Oct 20 00:00:00 UTC 2016
Laparoscopic Technique for Serial Collection of Para-Colonic, Left Colic, and Inferior Mesenteric Lymph Nodes in Macaques.
PloS one
Smedley J,Macalister R,Wangari S,Gathuka M,Ahrens J,Iwayama N,May D,Bratt D,O'Connor M,Munson P,Koday M,Lifson J,Fuller DH
14-7178 was used in Flow cytometry/Cell sorting to describe a minimally invasive laparoscopic technique.
Thu Jul 13 00:00:00 UTC 2017
COPA mutations impair ER-Golgi transport and cause hereditary autoimmune-mediated lung disease and arthritis.
Nature genetics
Watkin LB,Jessen B,Wiszniewski W,Vece TJ,Jan M,Sha Y,Thamsen M,Santos-Cortez RL,Lee K,Gambin T,Forbes LR,Law CS,Stray-Pedersen A,Cheng MH,Mace EM,Anderson MS,Liu D,Tang LF,Nicholas SK,Nahmod K,Makedonas G,Canter DL,Kwok PY,Hicks J,Jones KD,Penney S,Jhangiani SN,Rosenblum MD,Dell SD,Waterfield MR,Papa FR,Muzny DM,Zaitlen N,Leal SM,Gonzaga-Jauregui C,Boerwinkle E,Eissa NT,Gibbs RA,Lupski JR,Orange JS,Shum AK
14-7178 was used in Flow cytometry/Cell sorting to investigate how a vesicular transport protein and a syndrome of autoimmunity manifested by lung and joint disease have an unexpected molecular link.
Mon Jun 01 00:00:00 UTC 2015
Definition of a pool of epitopes that recapitulates the T cell reactivity against major house dust mite allergens.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology
Hinz D,Oseroff C,Pham J,Sidney J,Peters B,Sette A
14-7178 was used in an ELISpot assay to define an epitope mixture that recapitulates, and might substitute for, house dust mite extract in terms of detecting and characterising Th2 cell responses.
Thu Oct 01 00:00:00 UTC 2015
Fiber composite slices for multiplexed immunoassays.
Biomicrofluidics
Kim J,Bae S,Song S,Chung K,Kwon S
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Radioimmune Assays
Wed Jul 01 00:00:00 UTC 2015
Fiber composite slices for multiplexed immunoassays.
Biomicrofluidics
Kim J,Bae S,Song S,Chung K,Kwon S
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Radioimmune Assays
Wed Jul 01 00:00:00 UTC 2015
Differential requirement for P2X7R function in IL-17 dependent vs. IL-17 independent cellular immune responses.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons
Sullivan JA,Jankowska-Gan E,Shi L,Roenneburg D,Hegde S,Greenspan DS,Wilkes DS,Denlinger LC,Burlingham WJ
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Neutralization
Tue Jul 01 00:00:00 UTC 2014
Protein microarray analysis reveals BAFF-binding autoantibodies in systemic lupus erythematosus.
The Journal of clinical investigation
Price JV,Haddon DJ,Kemmer D,Delepine G,Mandelbaum G,Jarrell JA,Gupta R,Balboni I,Chakravarty EF,Sokolove J,Shum AK,Anderson MS,Cheng MH,Robinson WH,Browne SK,Holland SM,Baechler EC,Utz PJ
14-7178 was used in an ELISA assay to study how serum factor protein microarrays facilitate the detection of auto-antibody reactivity to serum factors in human samples.
Sun Dec 01 00:00:00 UTC 2013
Enhanced production of IL-17A in patients with severe asthma is inhibited by 1α,25-dihydroxyvitamin D3 in a glucocorticoid-independent fashion.
The Journal of allergy and clinical immunology
Nanzer AM,Chambers ES,Ryanna K,Richards DF,Black C,Timms PM,Martineau AR,Griffiths CJ,Corrigan CJ,Hawrylowicz CM
14-7178 was used in Flow cytometry/Cell sorting to investigate the production of the Th17-associated cytokines IL-17A and IL-22 by peripheral blood mononuclear cells from patients with moderate-to-severe asthma.
Thu Aug 01 00:00:00 UTC 2013
Human regulatory T cells do not suppress the antitumor immunity in the bone marrow: a role for bone marrow stromal cells in neutralizing regulatory T cells.
Clinical cancer research : an official journal of the American Association for Cancer Research
Guichelaar T,Emmelot ME,Rozemuller H,Martini B,Groen RW,Storm G,Lokhorst HM,Martens AC,Mutis T
14-7178 was used in Flow cytometry/Cell sorting to investigate Treg immunobiology and indicate the conditional benefits of future Treg-based terapies.
Fri Mar 15 00:00:00 UTC 2013
Gluten-sensitive enteropathy coincides with decreased capability of intestinal T cells to secrete IL-17 and IL-22 in a macaque model for celiac disease.
Clinical immunology (Orlando, Fla.)
Xu H,Feely SL,Wang X,Liu DX,Borda JT,Dufour J,Li W,Aye PP,Doxiadis GG,Khosla C,Veazey RS,Sestak K
14-7178 was used in Flow cytometry/Cell sorting to indicate that IL-17/22-producing cells play an important role in maintenance of intestinal mucosa in gluten-sensitive primates.
Mon Apr 01 00:00:00 UTC 2013
Rate of AIDS progression is associated with gastrointestinal dysfunction in simian immunodeficiency virus-infected pigtail macaques.
Journal of immunology (Baltimore, Md. : 1950)
Canary LA,Vinton CL,Morcock DR,Pierce JB,Estes JD,Brenchley JM,Klatt NR
14-7178 was used in Flow cytometry/Cell sorting to determine whether pre-existing GI-associated mucosal damage affects the progression of AIDS following HIV infection.
Fri Mar 15 00:00:00 UTC 2013
CCL20 Secretion from the Nucleus Pulposus Improves the Recruitment of CCR6-Expressing Th17 Cells to Degenerated IVD Tissues.
PloS one
Zhang W,Nie L,Wang Y,Wang XP,Zhao H,Dongol S,Maharjan S,Cheng L
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Fri Oct 13 00:00:00 UTC 2017
CCL20 Secretion from the Nucleus Pulposus Improves the Recruitment of CCR6-Expressing Th17 Cells to Degenerated IVD Tissues.
PloS one
Zhang W,Nie L,Wang Y,Wang XP,Zhao H,Dongol S,Maharjan S,Cheng L
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Flow Cytometry
Fri Oct 13 00:00:00 UTC 2017
Loss of mucosal CD103+ DCs and IL-17+ and IL-22+ lymphocytes is associated with mucosal damage in SIV infection.
Mucosal immunology
Klatt NR,Estes JD,Sun X,Ortiz AM,Barber JS,Harris LD,Cervasi B,Yokomizo LK,Pan L,Vinton CL,Tabb B,Canary LA,Dang Q,Hirsch VM,Alter G,Belkaid Y,Lifson JD,Silvestri G,Milner JD,Paiardini M,Haddad EK,Brenchley JM
14-7178 was used in Immunohistochemistry to study the underlying mechanisms responsible for the association of HIV/SIV infection with microbial translocation and persistent pathological immune activation.
Thu Nov 01 00:00:00 UTC 2012
IL-17-producing innate lymphoid cells are restricted to mucosal tissues and are depleted in SIV-infected macaques.
Mucosal immunology
Xu H,Wang X,Liu DX,Moroney-Rasmussen T,Lackner AA,Veazey RS
14-7178 was used in Flow cytometry/Cell sorting to indicate that ILC17 cells may be involved in innate mucosal immune responses, and their loss may contribute to loss of intestinal mucosal integrity.
Thu Nov 01 00:00:00 UTC 2012
Prolonged tenofovir treatment of macaques infected with K65R reverse transcriptase mutants of SIV results in the development of antiviral immune responses that control virus replication after drug withdrawal.
Retrovirology
Van Rompay KK,Trott KA,Jayashankar K,Geng Y,LaBranche CC,Johnson JA,Landucci G,Lipscomb J,Tarara RP,Canfield DR,Heneine W,Forthal DN,Montefiori D,Abel K
14-7178 was used in Flow cytometry/Cell sorting to provide hope for a functional cure of HIV infection via immunotherapeutic strategies that boost antiviral immunity and reduce the need for continuous antiretroviral therapy.
Tue Jul 17 00:00:00 UTC 2012
CD5 costimulation induces stable Th17 development by promoting IL-23R expression and sustained STAT3 activation.
Blood
de Wit J,Souwer Y,van Beelen AJ,de Groot R,Muller FJ,Klaasse Bos H,Jorritsma T,Kapsenberg ML,de Jong EC,van Ham SM
14-7178 was used in an ELISA assay to suggest that costimulation via CD5, rather than classic costimulation via CD28, primes naive T cells for stable Th17 development by promoting the expression of IL-23R.
Thu Dec 01 00:00:00 UTC 2011
Virus infection stages and distinct Th1 or Th17/Th22 T-cell responses in malaria/SHIV coinfection correlate with different outcomes of disease.
The Journal of infectious diseases
Ryan-Payseur B,Ali Z,Huang D,Chen CY,Yan L,Wang RC,Collins WE,Wang Y,Chen ZW
14-7178 was used in Flow cytometry/Cell sorting to suggest that virus infection status and selected Th1 or Th17/Th22 responses after malaria/AIDS-virus coinfection correlate with distinct outcomes of virus infection and malaria.
Tue Nov 01 00:00:00 UTC 2011
IL-1β and TGF-β act antagonistically in induction and differentially in propagation of human proinflammatory precursor CD4+ T cells.
Journal of immunology (Baltimore, Md. : 1950)
Hebel K,Rudolph M,Kosak B,Chang HD,Butzmann J,Brunner-Weinzierl MC
14-7178 was used in an ELISpot assay to monitor early T cell differentiation after T cell activation in the presence of different inflammatory cytokines.
Thu Dec 01 00:00:00 UTC 2011
Anticardiac myosin immunity and chronic allograft vasculopathy in heart transplant recipients.
Journal of immunology (Baltimore, Md. : 1950)
Kalache S,Dinavahi R,Pinney S,Mehrotra A,Cunningham MW,Heeger PS
14-7178 was used in an ELISpot assay to investigate the pathogenesis of chronic allograft vasculopathy in heart transplant failure.
Fri Jul 15 00:00:00 UTC 2011
Activated memory B cells may function as antigen-presenting cells in the joints of children with juvenile idiopathic arthritis.
Arthritis and rheumatism
Morbach H,Wiegering V,Richl P,Schwarz T,Suffa N,Eichhorn EM,Eyrich M,Girschick HJ
14-7178 was used in Flow cytometry/Cell sorting to investigate whether synovial B cells in juvenile idiopathic arthritis display features of antigen-presenting cells.
Tue Nov 01 00:00:00 UTC 2011
Licensing virus-specific T cells to secrete the neutrophil attracting chemokine CXCL-8 during hepatitis B virus infection.
PloS one
Gehring AJ,Koh S,Chia A,Paramasivam K,Chew VS,Ho ZZ,Lee KH,Maini MK,Madhavan K,Lim SG,Bertoletti A
14-7178 was used in an ELISA assay to investigate whether during different phases of HBV infection, virus-specific T-cells can acquire specific proinflammatory functions.
Fri Dec 30 00:00:00 UTC 2011
Molecular determinants of T cell epitope recognition to the common Timothy grass allergen.
Journal of immunology (Baltimore, Md. : 1950)
Oseroff C,Sidney J,Kotturi MF,Kolla R,Alam R,Broide DH,Wasserman SI,Weiskopf D,McKinney DM,Chung JL,Petersen A,Grey H,Peters B,Sette A
14-7178 was used in an ELISpot assay to identify that intrinsic features of the allergen protein might influence immunogenicity at the level of T cell reactivity.
Thu Jul 15 00:00:00 UTC 2010
A comprehensive ex vivo functional analysis of human NKT cells reveals production of MIP1-α and MIP1-β, a lack of IL-17, and a Th1-bias in males.
PloS one
Snyder-Cappione JE,Tincati C,Eccles-James IG,Cappione AJ,Ndhlovu LC,Koth LL,Nixon DF
14-7178 was used in an ELISpot assay to define the scope and magnitude of the ex vivo functional capacity of human natural killer T cells and further assess for gender-specific differences.
Wed Nov 03 00:00:00 UTC 2010
CD49d provides access to "untouched" human Foxp3+ Treg free of contaminating effector cells.
Blood
Kleinewietfeld M,Starke M,Di Mitri D,Borsellino G,Battistini L,Rötzschke O,Falk K
14-7178 was used in Flow cytometry/Cell sorting to investigate more effective markers for the isolation of Tregs for their use in adoptive transfer.
Thu Jan 22 00:00:00 UTC 2009
A monoclonal antibody selection for immunohistochemical examination of lymphoid tissues from non-human primates.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Kap YS,van Meurs M,van Driel N,Koopman G,Melief MJ,Brok HP,Laman JD,'t Hart BA
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Immunohistochemistry
Tue Dec 01 00:00:00 UTC 2009
Interleukin-17 stimulates C-reactive protein expression in hepatocytes and smooth muscle cells via p38 MAPK and ERK1/2-dependent NF-kappaB and C/EBPbeta activation.
The Journal of biological chemistry
Patel DN,King CA,Bailey SR,Holt JW,Venkatachalam K,Agrawal A,Valente AJ,Chandrasekar B
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Functional assay
Fri Sep 14 00:00:00 UTC 2007
Interleukin-17 stimulates C-reactive protein expression in hepatocytes and smooth muscle cells via p38 MAPK and ERK1/2-dependent NF-kappaB and C/EBPbeta activation.
The Journal of biological chemistry
Patel DN,King CA,Bailey SR,Holt JW,Venkatachalam K,Agrawal A,Valente AJ,Chandrasekar B
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Functional assay
Fri Sep 14 00:00:00 UTC 2007
Interleukin-17 stimulates C-reactive protein expression in hepatocytes and smooth muscle cells via p38 MAPK and ERK1/2-dependent NF-kappaB and C/EBPbeta activation.
The Journal of biological chemistry
Patel DN,King CA,Bailey SR,Holt JW,Venkatachalam K,Agrawal A,Valente AJ,Chandrasekar B
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Functional assay
Fri Sep 14 00:00:00 UTC 2007
Interleukin-17 stimulates C-reactive protein expression in hepatocytes and smooth muscle cells via p38 MAPK and ERK1/2-dependent NF-kappaB and C/EBPbeta activation.
The Journal of biological chemistry
Patel DN,King CA,Bailey SR,Holt JW,Venkatachalam K,Agrawal A,Valente AJ,Chandrasekar B
Published figure using IL-17A monoclonal antibody (Product # 14-7178-81) in Functional assay
Fri Sep 14 00:00:00 UTC 2007