Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Flow Cytometry (Flow) |
0.50 µg/test | View 1 publication 1 publication |
Inhibition Assays (IA) |
- | View 1 publication 1 publication |
Product Specifications | |
---|---|
Species Reactivity |
Mouse |
Published species |
Mouse |
Host/Isotype |
Rat / IgG1, kappa |
Recommended Isotype Control |
Rat IgG1 kappa Isotype Control (eBRG1), Alexa Fluor™ 488, eBioscience™ |
Class |
Monoclonal |
Type |
Antibody |
Clone |
eBioTC11-18H10.1 |
Conjugate |
Alexa Fluor™ 488 |
Excitation/Emission Profile |
View spectra |
Form |
Liquid |
Concentration |
0.5 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.2 |
Contains |
0.09% sodium azide |
Storage conditions |
4° C, store in dark, DO NOT FREEZE! |
RRID |
AB_493960 |
Description: The eBioTC11-18H10.1 antibody reacts with mouse IL-17A. The eBioTC11-18H10.1 antibody is a neutralizing antibody. Interleukin-17A (IL-17A) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. The cDNA encoding human IL-17A was isolated from a library of CD4+ T cells; the encoded protein exhibits 72 percent amino acid identity with HVS13 , an open reading frame from a T lymphotropic Herpesvirus saimiri, and 63 percent with mouse CTLA-8 (cytotoxic T-lymphocyte associated antigen-8). Human IL-17A exists as glycosylated 20-30 kD homodimers. High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells. IL-17A enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. Human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse IL-17A receptor.
IL-23-dependent, IL-17A-producing CD4+ T cells (Th-17 cells) have been identified as a unique subset of Th cells that develops along a pathway that is distinct from the Th1- and Th2- cell differentiation pathways. The hallmark effector molecules of Th1 and Th2 cells, e.g., IFN-gamma and IL-4, have each been found to negatively regulate the generation of these Th-17 cells.
Applications Reported: The eBioTC11-18H10.1 antibody has been reported for use as the capture antibody in mouse IL-17A ELISA and ELISPOT assay, for neutralization of IL-17A bioactivity, and for intracellular staining of IL-17A-producing cells for flow cytometric analysis.
Applications Tested: The fluorochrome-conjugated eBioTC11-18H10.1 antibody has been tested by intracellular staining for flow cytometric analysis and can be used at less than or equal to 0.50 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: CTLA-8; Cytotoxic T-lymphocyte-associated antigen 8; il 17; IL-17; ILN; Interleukin; interleukin 17; Interleukin-17A; Interleukin17A; R-IL-17-A
Gene Aliases: Ctla-8; Ctla8; IL-17; IL-17A; Il17; Il17a
UniProt ID: (Mouse) Q62386
Entrez Gene ID: (Mouse) 16171
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support