Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description: BMS1042 (7G8) recognises natural and recombinant porcine IL-18. The antibody does not recognise porcine IL-1beta, IL-8, IL-12 or IFN-gamma, nor does it cross react with human or murine IL-18.
Applications Tested: ELISA, Western Blotting.
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytokine; IFN-gamma-inducing factor; il 18; IL-1 gamma; IL-18; ILN; Interferon gamma-inducing factor; Interleukin; interleukin 18 (interferon-gamma-inducing factor); Interleukin-1 gamma; Interleukin-18; Interleukin18; MGC12320
Gene Aliases: IGIF; IL-18; IL18
UniProt ID: (Pig) O19073
Entrez Gene ID: (Pig) 397057
Molecular Function: cytokine interleukin superfamily intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support