Product References
Dysregulated Peripheral Invariant Natural Killer T Cells in Plaque Psoriasis Patients.
Frontiers in cell and developmental biology
Hu Y,Chen Y,Chen Z,Zhang X,Guo C,Yu Z,Xu P,Sun L,Zhou X,Gong Y,Yu Q,Shi Y
Published figure using IL-4 monoclonal antibody (Product # 12-7049-42) in Flow Cytometry
Wed Feb 23 00:00:00 EST 2022
Cord Blood T Cells Expressing High and Low PKCζ Levels Develop into Cells with a Propensity to Display Th1 and Th9 Cytokine Profiles, Respectively.
International journal of molecular sciences
Perveen K,Quach A,McPhee A,Prescott SL,Barry SC,Hii CS,Ferrante A
12-7049-42 was used in Flow Cytometry to support the view that low CBTC PKCζ levels relate to the increased risk of developing allergic diseases.
Wed May 05 00:00:00 EDT 2021
Cord Blood T Cells Expressing High and Low PKCζ Levels Develop into Cells with a Propensity to Display Th1 and Th9 Cytokine Profiles, Respectively.
International journal of molecular sciences
Perveen K,Quach A,McPhee A,Prescott SL,Barry SC,Hii CS,Ferrante A
12-7049-42 was used in Flow Cytometry to support the view that low CBTC PKCζ levels relate to the increased risk of developing allergic diseases.
Wed May 05 00:00:00 EDT 2021
Recombinant human IL-37 inhibited endometriosis development in a mouse model through increasing Th1/Th2 ratio by inducing the maturation of dendritic cells.
Reproductive biology and endocrinology : RB&E
Li L,Liao Z,Ye M,Jiang J
12-7049-42 was used in Flow cytometry/Cell sorting to study recombinant human IL-37 (rhIL-37) is an inhibitor of inflammation.
Tue Aug 24 00:00:00 EDT 2021
Single-cell multi-omics analysis presents the landscape of peripheral blood T-cell subsets in human chronic prostatitis/chronic pelvic pain syndrome.
Journal of cellular and molecular medicine
Zhang M,Liu Y,Chen J,Chen L,Meng J,Yang C,Yin S,Zhang X,Zhang L,Hao Z,Chen X,Liang C
12-7049 was used in Flow cytometry/Cell sorting to digitally quantify both the protein and mRNA expression levels in single peripheral blood T cells from two chronic prostatitis / chronic pelvic pain patients and two healthy controls.
Tue Dec 01 00:00:00 EST 2020
Standardized 11-color flow cytometry panel for the functional phenotyping of human T regulatory cells.
Journal of biological methods
Manuszak C,Brainard M,Thrash E,Hodi FS,Severgnini M
Published figure using IL-4 monoclonal antibody (Product # 12-7049-42) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Th2 Biased Immunity With Altered B Cell Profiles in Circulation of Patients With Sporotrichosis Caused by Sporothrix globosa.
Frontiers in immunology
Zu J,Yao L,Song Y,Cui Y,Guan M,Chen R,Zhen Y,Li S
Published figure using IL-4 monoclonal antibody (Product # 12-7049-42) in Flow Cytometry
Mon May 31 00:00:00 EDT 2021
Platelet-Derived GARP Induces Peripheral Regulatory T Cells-Potential Impact on T Cell Suppression in Patients with Melanoma-Associated Thrombocytosis.
Cancers
Zimmer N,Krebs FK,Zimmer S,Mitzel-Rink H,Kumm EJ,Jurk K,Grabbe S,Loquai C,Tuettenberg A
Published figure using IL-4 monoclonal antibody (Product # 12-7049-42) in Flow Cytometry
Sat Dec 05 00:00:00 EST 2020
CD4+ Th immunogenicity of the Ascaris spp. secreted products.
NPJ vaccines
Ebner F,Morrison E,Bertazzon M,Midha A,Hartmann S,Freund C,Álvaro-Benito M
12-7049-42 was used in Flow Cytometry to identify the first human T cell epitope against Ascaris spp. and represents an easily adaptable platform for characterization of complex antigens.
Thu Mar 18 00:00:00 EDT 2021
CD4 T cell loss and Th2 and Th17 bias are associated with the severity of severe fever with thrombocytopenia syndrome (SFTS).
Clinical immunology (Orlando, Fla.)
Li MM,Zhang WJ,Weng XF,Li MY,Liu J,Xiong Y,Xiong SE,Zou CC,Wang H,Lu MJ,Yang DL,Peng C,Zheng X
Published figure using IL-4 monoclonal antibody (Product # 12-7049-42) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
CD4 T cell loss and Th2 and Th17 bias are associated with the severity of severe fever with thrombocytopenia syndrome (SFTS).
Clinical immunology (Orlando, Fla.)
Li MM,Zhang WJ,Weng XF,Li MY,Liu J,Xiong Y,Xiong SE,Zou CC,Wang H,Lu MJ,Yang DL,Peng C,Zheng X
Published figure using IL-4 monoclonal antibody (Product # 12-7049-42) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
Lactobacillus rhamnosus GG-Derived Soluble Mediators Modulate Adaptive Immune Cells.
Frontiers in immunology
Ludwig IS,Broere F,Manurung S,Lambers TT,van der Zee R,van Eden W
12-7049 was used in Flow cytometry/Cell sorting to determine the role of Lactobacillus rhamosus soluble mediators in adaptive immune responses.
Tue Sep 26 00:00:00 EDT 2023
Genetic Architecture of Adaptive Immune System Identifies Key Immune Regulators.
Cell reports
Lagou V,Garcia-Perez JE,Smets I,Van Horebeek L,Vandebergh M,Chen L,Mallants K,Prezzemolo T,Hilven K,Humblet-Baron S,Moisse M,Van Damme P,Boeckxstaens G,Bowness P,Dubois B,Dooley J,Liston A,Goris A
12-7049-42 was used in Flow Cytometry to identify eight genome-wide significant associations explaining 6%-20% of variance in immune system regulation.
Tue Oct 16 00:00:00 EDT 2018
Functionalization-dependent effects of cellulose nanofibrils on tolerogenic mechanisms of human dendritic cells.
International journal of nanomedicine
Tomić S,Ilić N,Kokol V,Gruden-Movsesijan A,Mihajlović D,Bekić M,Sofronić-Milosavljević L,Čolić M,Vučević D
12-7049 was used in Flow cytometry/Cell sorting to investigate how differentially functionalised cellulose nanofibrils affect dendritic cell properties, their viability, morphology, differentiation and maturation potential, and the capacity to regulate T cell-mediated immune response.
Mon Dec 31 00:00:00 EST 2018
Impact of Enhancer of Zeste Homolog 2 on T Helper Cell-Mediated Allergic Rhinitis.
Frontiers in immunology
Hou TY,Chen MR,Chou YC,Kan PC,Tsai YT,Cha TL
12-7049 was used in Flow cytometry/Cell sorting to investigate the function of enhancer of zeste homolog 2 in human Th1 and Th2 cell differentiation, and its association with allergic rhinitis.
Wed Nov 20 00:00:00 EST 2019
Impact of Enhancer of Zeste Homolog 2 on T Helper Cell-Mediated Allergic Rhinitis.
Frontiers in immunology
Hou TY,Chen MR,Chou YC,Kan PC,Tsai YT,Cha TL
12-7049 was used in Flow cytometry/Cell sorting to investigate the function of enhancer of zeste homolog 2 in human Th1 and Th2 cell differentiation, and its association with allergic rhinitis.
Wed Nov 20 00:00:00 EST 2019
Mucosal and systemic immune modulation by Trichuris trichiura in a self-infected individual.
Parasite immunology
Dige A,Rasmussen TK,Nejsum P,Hagemann-Madsen R,Williams AR,Agnholt J,Dahlerup JF,Hvas CL
12-7049-42 was used in Flow cytometry/Cell sorting to investigated how self-infection with Trichuris (T.) trichiura in an adult man without intestinal disease affected mucosal and systemic immunity.
Sun Jan 01 00:00:00 EST 2017
A pilot study showing associations between frequency of CD4(+) memory cell subsets at diagnosis and duration of partial remission in type 1 diabetes.
Clinical immunology (Orlando, Fla.)
Moya R,Robertson HK,Payne D,Narsale A,Koziol J,Davies JD
12-7049 was used in Flow cytometry/Cell sorting to assess whether frequencies of CD4(+) T cell subsets in children newly diagnosed with type 1 diabetes are associated with length of partial remission.
Sun May 01 00:00:00 EDT 2016
CD16+ Monocyte Subset Was Enriched and Functionally Exacerbated in Driving T-Cell Activation and B-Cell Response in Systemic Lupus Erythematosus.
Frontiers in immunology
Zhu H,Hu F,Sun X,Zhang X,Zhu L,Liu X,Li X,Xu L,Shi L,Gan Y,Su Y
Published figure using IL-4 monoclonal antibody (Product # 12-7049-42) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
Effects of transcutaneous acupoint electrical stimulation on the imbalance of Th1, Th2, Th17 and Treg cells following thoracotomy of patients with lung cancer.
Experimental and therapeutic medicine
Wu H,Wang K,Li G,Meng D,Han J,Wang G,Li YU
12-7049 was used in Flow cytometry/Cell sorting to demonstrate that transcutaneous acupoint electrical stimulation was able to partially attenuate the postoperative immune depression of patients with lung cancer.
Mon Feb 01 00:00:00 EST 2016
A SnoRNA-derived piRNA interacts with human interleukin-4 pre-mRNA and induces its decay in nuclear exosomes.
Nucleic acids research
Zhong F,Zhou N,Wu K,Guo Y,Tan W,Zhang H,Zhang X,Geng G,Pan T,Luo H,Zhang Y,Xu Z,Liu J,Liu B,Gao W,Liu C,Ren L,Li J,Zhou J,Zhang H
Published figure using IL-4 monoclonal antibody (Product # 12-7049-42) in Flow Cytometry
Wed Dec 02 00:00:00 EST 2015
T-helper 17 cell polarization in pulmonary arterial hypertension.
Chest
Hautefort A,Girerd B,Montani D,Cohen-Kaminsky S,Price L,Lambrecht BN,Humbert M,Perros F
12-7049 was used in Flow cytometry/Cell sorting to study T helper 17 cell immune polarization in patients with pulmonary arterial hypertension.
Mon Jun 01 00:00:00 EDT 2015
Do patients with a failed metal-on-metal hip implant with a pseudotumor present differences in their peripheral blood lymphocyte subpopulations?
Clinical orthopaedics and related research
Catelas I,Lehoux EA,Hurda I,Baskey SJ,Gala L,Foster R,Kim PR,Beaulé PE
12-7049 was used in Flow cytometry/Cell sorting to reveal the presence of a type IV hypersensitivity reaction in patients with a failed hip implant with a pseudotumour.
Tue Dec 01 00:00:00 EST 2015
Depressive symptoms post hip fracture in older adults are associated with phenotypic and functional alterations in T cells.
Immunity & ageing : I & A
Duggal NA,Upton J,Phillips AC,Hampson P,Lord JM
12-7049 was used in Flow cytometry/Cell sorting to investigate the synergistic effects of hip fracture physical trauma and psychological depressive symptoms on the aged immune system.
Wed Jan 28 00:00:00 EST 2015
BET bromodomain inhibition suppresses TH17-mediated pathology.
The Journal of experimental medicine
Mele DA,Salmeron A,Ghosh S,Huang HR,Bryant BM,Lora JM
12-7049 was used in Flow cytometry/Cell sorting to identify BET proteins as a fundamental link between chromatin signalling and T(H)17 biology.
Mon Oct 21 00:00:00 EDT 2013
Brefeldin A, but not monensin, enables flow cytometric detection of interleukin-4 within peripheral T cells responding to ex vivo stimulation with Chlamydia trachomatis.
Journal of immunological methods
Vicetti Miguel RD,Maryak SA,Cherpes TL
12-7049 was used in Flow cytometry/Cell sorting to compare the ability of brefeldin A and monensin to concurrently trap 4 cytokines within peripheral blood mononuclear cells.
Wed Oct 31 00:00:00 EDT 2012