Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Mitofilin is a transmembrane protein of the inner mitochondrial membrane and it is associated with a large multimeric protein complex of about 1200 kDa. Mitofilin has critical functions in mitochondrial morphology and mitochondrial fusion and fission, specifically in the formation of tubular cristae and cristae junctions. Abnormal mitochondrial morphology has been implicated in many human diseases such as myopathy, cardiomyopathy, rhabdomyosarcoma and Whartin's tumor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell proliferation-inducing gene 4/52 protein; cell proliferation-inducing protein 52; IMMT antibody; MICOS complex subunit MIC60; mitochondrial inner membrane organizing system 2; Mitochondrial inner membrane protein; Mitofilin; motor protein; p87/89; p87/89 antibody; proliferation-inducing gene 4
Gene Aliases: HMP; IMMT; MIC60; MINOS2; P87; P87/89; P89; PIG4; PIG52
UniProt ID: (Human) Q16891
Entrez Gene ID: (Human) 10989
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support