Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Immunohistochemistry (IHC) |
1:50-1:250 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Between 1 and 50 |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
0.10 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
TBS with 0.1% BSA |
Contains |
0.09% sodium azide |
Storage conditions |
4° C |
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
This gene encodes a member of the IQGAP family. The protein contains four IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. Expression of the protein is upregulated by gene amplification in two gastric cancer cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DISP2; DISPB; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs
Gene Aliases: HUMORFA01; IQGAP1; KIAA0051; p195; SAR1
UniProt ID: (Human) P46940
Entrez Gene ID: (Human) 8826
Molecular Function:
G-protein modulator
protein-binding activity modulator
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support