Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
Mouse anti Human Integrin alpha 9 beta 1 antibody, clone Y9A2 recognizes the human alpha9 integrin, a approximately 140 kDa glycoprotein expressed as a heterodimer in association with the beta1 integrin.
Integrins are heterodimeric proteins made up of alpha and beta subunits. At least 18 alpha and 8 beta subunits have been described in mammals. Integrin family members are membrane receptors involved in cell adhesion and recognition in a variety of processes including embryogenesis, hemostasis, tissue repair, immune response and metastatic diffusion of tumor cells. This gene encodes a beta subunit. Multiple alternatively spliced transcript variants which encode different protein isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD29; Fibronectin receptor subunit beta; Glycoprotein IIa; GPIIA; integrin alpha 9; Integrin alpha-9; Integrin alpha-RLC; integrin beta 1; Integrin beta-1; integrin beta-1 subunit; Integrin beta1; integrin VLA-4 beta subunit; integrin, alpha 9; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); very late activation protein, beta polypeptide; VLA-4 subunit beta
Gene Aliases: ALPHA-RLC; CD29; FNRB; GPIIA; ITGA4L; ITGA9; ITGB1; MDF2; MSK12; RLC; VLA-BETA; VLAB
UniProt ID: (Human) Q13797, (Pig) Q9GLP0, (Human) P05556
Entrez Gene ID: (Pig) 100626707, (Human) 3680, (Pig) 397019, (Human) 3688, (Guinea pig) 100716659, (Guinea pig) 100719502
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support