Search Thermo Fisher Scientific
PA1-27326 detects Influenza A Virus M1 Protein from virus samples. PA1-27326 will not react with Influenza B, Adenovirus, Respiratory syncytial virus, or Parainfluenza viruses(1-3).
PA1-27326 has been successfully used in ELISA and immunofluorescence procedures. It is unsuitable for use in immunohistochemistry.
The PA1-27326 immunogen is Influenza A strain Phillipines (H3N2).
Store short term at 4C. Store at -20C long term avoiding freeze-thaw cycles.
Influenza A virus matrix 1, M1, is a critical protein required for assembly and budding. Hemagglutinin associates with M1 via its cytoplasmic tail and transmembrane domain. The M2 and nucleoprotein proteins are critical in the replication cycle of influenza viruses. The M2 channel protein is an essential component of the viral envelope because of its ability to form a highly selective, pH-regulated, proton-conducting channel. The M2 channel allows protons to enter the interior of the virus and acidification weakens the interaction of the M1 protein with the ribonuclear core.
Protein Aliases: Influenza M1