Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGDMGDPPKK KRLISLCVGC GNQIHDQYIL RVSPDLEWHA ACLKCAECNQ YLDESCTCFV RDGKTYCKRD YIRLYGIKCA KCSIGFSKND FVMRARSKVY HIECFRCVAC SRQLIPGDEF ALREDGLFCR ADHDVVERAS LGAGDPLSPL HPARPLQMAA EPISARQPAL RPHVHKQPEK TTRVRTVLNE KQLHTLRTCY AANPRPDALM KEQLVEMTGL SPRVIRVWFQ NKRCKDKKRS IMMKQLQQQQ PNDKTNIQGM TGTPMVAASP ERHDGGLQAN PVEVQSYQPP WKVLSDFALQ SDIDQPAFQQ LVNFSEGGPG SNSTGSEVAS MSSQLPDTPN SMVASPIEA
This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Insulin gene enhancer protein ISL-1; ISL1 transcription factor, LIM/homeodomain; Islet-1
Gene Aliases: Isl-1; ISL1; ISLET1
UniProt ID: (Human) P61371
Entrez Gene ID: (Human) 3670
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support