Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MSRSKRDNN FYSVEIGDST FTVLKRYQNL KPIGSGAQGI VCAAYDAILE RNVAIKKLSR PFQNQTHAKR AYRELVLMKC VNHKNIIGLL NVFTPQKSLE EFQDVYIVME LMDANLCQVI QMELDHERMS YLLYQMLCGI KHLHSAGIIH RDLKPSNIVV KSDCTLKILD FGLARTAGTS FMMTPYVVTR YYRAPEVILG MGYKENVDIW SVGCIMGEMI KGGVLFPGTD HIDQWNKVIE QLGTPCPEFM KKLQPTVRTY VENRPKYAGY SFEKLFPDVL FPADSEHNKL KASQARDLLS KMLVIDASKR ISVDEALQHP YINVWYDPSE AEAPPPKIPD KQLDEREHTI EEWKELIYKE VMDLEERTKN GVIRGQPSPL AQVQQ
MAPK8 (JNK1) is activated by the presence of misfolded proteins in the endoplasmic reticulum. It is also a crucial mediator of insulin resistance and obesity. It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Jun N-terminal kinase 1; FLJ12099; FLJ33785; JNK-46; JNK1 beta1 protein kinase; JNK3; JNK3A; JUN N-terminal kinase; MAP kinase 8; MAPK 10; MAPK 8; MAPK10; MGC50974; mitogen activated protein kinase 8; Mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; MK10; OTTHUMP00000019552; p493F12; p54 gamma; p54bSAPK; Phospho-JNK; PRKM10; protein kinase JNK1; protein kinase mitogen-activated 8; SAPK gamma; SAPK1c; stress-activated protein kinase 1; Stress-activated protein kinase 1c; Stress-activated protein kinase JNK1
Gene Aliases: AI849689; JNK; JNK-46; JNK1; JNK1A2; JNK21B1/2; MAPK8; PRKM8; SAPK1; SAPK1C
UniProt ID: (Human) P45983, (Rat) P49185, (Mouse) Q91Y86
Entrez Gene ID: (Human) 5599, (Rat) 116554, (Mouse) 26419
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support