Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For maximum recovery of product, centrifuge the vial prior to removing the cap.
KChIP2b is an isoform of the Kv Channel-Interacting Protein 2 (KChIP2), which is a regulatory subunit that interacts with voltage-gated potassium (Kv) channels. This isoform modulates the biophysical properties of Kv channels, including their inactivation kinetics and surface expression, thereby influencing cardiac and neuronal excitability. Dysregulation of KChIP2b can impact heart function and has been implicated in conditions such as cardiac arrhythmias.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A-type potassium channel modulatory protein 2; A-type potassium channel modulatory protein KCNIP2; Cardiac voltage-gated potassium channel modulatory subunit; KChIP2; Kv channel interacting protein 2; Kv channel-interacting protein; Kv channel-interacting protein 2; potassium channel auxiliary subunit KCHIP2; Potassium channel-interacting protein 2
Gene Aliases: KCHIP2; KCNIP2
UniProt ID: (Human) Q9NS61, (Mouse) Q9JJ69, (Rat) Q9JM59
Entrez Gene ID: (Pig) 100037948, (Human) 30819, (Dog) 403598, (Mouse) 80906, (Rat) 56817
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support