Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KDEL Antibodies

          QED Bioscience

          KDEL Receptor Monoclonal Antibody (KR-10)

          View all (22) KDEL antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite KDEL Receptor Monoclonal Antibody (KR-10)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          KDEL Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          KDEL Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KDEL Receptor Antibody (11078-100UG) in ICC/IF

          Immunocytochemistry/Immunofluorescence analysis using Mouse Anti-KDEL Receptor Monoclonal Antibody, Clone KR-10 (11078). Tissue: NRK cells. Species: Rat. Primary Antibody: Mouse Anti-KDEL Receptor Monoclonal Antibody (11078) at 1:1000. Secondary Antibody: APC Goat Anti-Mouse (red). Counterstain: DAPI (blue) nuclear stain. KR-10 staining red; DAPI staining blue. Merged images. Courtesy of: Institute of Mol. ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KDEL Receptor Antibody in Immunocytochemistry (ICC/IF)
          KDEL Receptor Antibody in Western Blot (WB)
          KDEL Receptor Monoclonal Antibody (KR-10)

          Product Details

          11078-100UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, Chicken, Fruit fly, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat, Xenopus

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          KR-10

          Immunogen

          Peptide corresponding to aa 192-212 of bovine KDEL receptor
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.2, with 50% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          2-8°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble endoplasmic reticulum (ER) resident proteins and some membrane proteins. 78 and 94 kDa glucose regulated proteins, GRP78 and GRP94 respectively, and protein disulfide isomerase (PDI) all share the C-terminal KDEL sequence. The presence of carboxy-terminal KDEL appears to be necessary for ER retention and appears to be sufficient to reduce the secretion of proteins from the ER. This retention is reported to be mediated by a KDEL receptor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 108K heat shock protein; 108K HSP; 78 kDa glucose-regulated protein; 78-kD glucose-regulated protein precursor; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250|UniProtKB:P08113}; 98 kDa protein kinase; Binding-immunoglobulin protein; BiP; BiP protein; CG5183-PA; CG5183-PB; CG5183-PC; chaperone; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; Endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250|UniProtKB:P08113}; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; epididymis secretory sperm binding protein Li 89n; ER lumen protein retaining receptor 1; ER lumen protein retaining receptor 2; ER lumen protein-retaining receptor; ER lumen protein-retaining receptor 1; ERp99; glucose regulated protein, 78 kDa; glucose-regulated protein; glucose-regulated protein (grp78); glucose-regulated protein GRP94; glucose-regulated protein, 78kDa; gp96 homolog; GRP 78; GRP-78; GRP-94; GRP94; Heat shock 108 kDa protein; heat shock 70 kDa protein 5; heat shock 70 kDa protein 5a; heat shock 70kD protein 5; heat shock 70kD protein 5 (glucose-regulated protein, 78kD); heat shock 70kDa protein 5 (glucose-regulated protein); heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); heat shock protein 108; heat shock protein 108.; heat shock protein 5; Heat shock protein 70 family protein 5; Heat shock protein 90 kDa beta member 1; heat shock protein 90 kDa beta member 1 {ECO:0000250|UniProtKB:P08113}; heat shock protein 90, beta (Grp94), member 1; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; heat shock protein 90kDa beta, member 1; Heat shock protein family A member 5; Heat shock protein family C member 4; heavy-chain binding protein BiP; HSP 108; hsp108; hsp70; HSP70 family protein 5; HSP90 beta; hsp90b1 {ECO:0000250|UniProtKB:P08113}; immunoglobulin binding protein; Immunoglobulin heavy chain-binding protein; KDEL; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 1; KDEL receptor 1; KDEL receptor-like protein; KDEL-receptor; KdelR-PA; KdelR-PB; KdelR-PC; lethal (2) 31Em; Lys-Asp-Glu-Leu; Polymorphic tumor rejection antigen 1; PPK 98; Putative MAPK-activating protein PM23; RP23-446N16.1; Steroidogenesis-activator polypeptide; stress-inducible tumor rejection antigen gp96; Transferrin-binding protein; transforming growth factor alpha regulated gene 2; tumor rejection antigen (gp96) 1; Tumor rejection antigen 1; Tumor rejection antigen gp96; unnamed protein product; XAP-1 antigen

          View more View less

          Gene Aliases: 8030486F04Rik; AL022860; AU019543; AW215843; baffled; BEST:CK00230; BIP; BiP/Grp78; CG5183; CK00230; D2Wsu141e; D2Wsu17e; Dmel\CG5183; Dmel_CG5183; dmErd2; ECGP; endoplasmin; ERD2; ERD2.1; ERp99; GP96; GRP78; GRP94; HDEL; HEL-S-125m; HEL-S-89n; HEL35; Hsce70; hsp108; HSP90B1; HSPA5; hspa5a; HSPC4; I79_019946; I79_020931; KDELR; KDELR1; l(2)31Em; l(2)k00311; mBiP; PM23; ppk98; SEZ-7; Sez7; TA-3; Targ2; Tra-1; TRA1; XAP-1; XELAEV_180384511mg

          View more View less

          UniProt ID: (Pig) Q29092, (Rabbit) O18750, (Human) P14625, (Dog) P41148, (Bovine) Q95M18, (Chicken) P08110, (Chicken) Q90593, (Human) P11021, (Bovine) Q0VCX2, (Rat) Q66HD0, (Mouse) P08113, (Mouse) P20029, (Chinese hamster) G3I8R9, (Rat) P06761, (Bovine) P33946, (Human) P24390, (Fruit fly) O76767, (Mouse) Q99JH8, (Rat) Q569A6, (Xenopus) Q91883

          View more View less

          Entrez Gene ID: (Pig) 397191, (Rabbit) 100009456, (Human) 7184, (Dog) 404019, (Bovine) 282646, (Chicken) 374163, (Sheep) 101111703, (Chicken) 396487, (Sheep) 780447, (Dog) 480726, (Pig) 407060, (Human) 3309, (Rabbit) 100008764, (Bovine) 415113, (Chinese hamster) 100773331, (Rat) 362862, (Mouse) 22027, (Mouse) 14828, (Chinese hamster) 100689305, (Rat) 25617, (Pig) 100520962, (Bovine) 618184, (Sheep) 101122047, (Human) 10945, (Dog) 484406, (Rabbit) 100349892, (Fruit fly) 34427, (Chinese hamster) 100750912, (Mouse) 68137, (Rat) 361577, (Xenopus) 397850

          View more View less

          Function(s)
          ATP binding nucleotide binding RNA binding protein phosphatase inhibitor activity calcium ion binding protein binding hydrolase activity ATPase activity protein phosphatase binding protein binding involved in protein folding low-density lipoprotein particle receptor binding unfolded protein binding ATP-dependent protein folding chaperone heat shock protein binding misfolded protein binding enzyme binding protein domain specific binding protein serine/threonine kinase inhibitor activity ubiquitin protein ligase binding ribosome binding cadherin binding chaperone binding virion binding glycoprotein binding KDEL sequence binding ER retention sequence binding molecular_function Hsp90 family chaperone membrane trafficking regulatory protein
          Process(es)
          ER-associated ubiquitin-dependent protein catabolic process response to hypoxia protein folding protein transport positive regulation of Wnt signaling pathway retrograde protein transport, ER to cytosol actin rod assembly positive regulation of toll-like receptor signaling pathway protein folding in endoplasmic reticulum response to endoplasmic reticulum stress ERAD pathway negative regulation of apoptotic process sequestering of calcium ion cellular response to manganese ion cellular response to ATP protein localization to plasma membrane regulation of phosphoprotein phosphatase activity intracellular sequestering of iron ion response to unfolded protein endoplasmic reticulum unfolded protein response negative regulation of protein complex assembly cellular response to unfolded protein protein refolding chaperone mediated protein folding requiring cofactor negative regulation of IRE1-mediated unfolded protein response positive regulation of cell migration posttranslational protein targeting to membrane, translocation maintenance of protein localization in endoplasmic reticulum translational initiation ER overload response cerebellum structural organization cerebellar Purkinje cell layer development substantia nigra development negative regulation of transforming growth factor beta receptor signaling pathway positive regulation of protein ubiquitination IRE1-mediated unfolded protein response PERK-mediated unfolded protein response cellular response to glucose starvation positive regulation of transcription from RNA polymerase II promoter proteolysis involved in cellular protein catabolic process regulation of protein folding in endoplasmic reticulum cellular response to interleukin-4 regulation of ATF6-mediated unfolded protein response regulation of IRE1-mediated unfolded protein response regulation of PERK-mediated unfolded protein response negative regulation of PERK-mediated unfolded protein response toxin transport response to stress activation of signaling protein activity involved in unfolded protein response positive regulation of embryonic development cellular response to antibiotic T cell cytokine production retrograde vesicle-mediated transport, Golgi to ER T cell differentiation T cell apoptotic process protein retention in ER lumen vesicle-mediated transport ER to Golgi vesicle-mediated transport transport biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0