Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • KDEL Antibodies

          Rockland

          KDEL Receptor Monoclonal Antibody (KR-10)

          View all (21) KDEL antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite KDEL Receptor Monoclonal Antibody (KR-10)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          KDEL Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          KDEL Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KDEL Receptor Antibody (500-301-F86) in ICC/IF

          Immunofluorescence of mouse anti-KDEL antibody. Tissue: NRK cells. Antigen retrieval: not required. Primary Antibody: KDEL at 1 µg/mL for 1 hr at RT. Secondary antibody: Fluorescein Anti-Mouse secondary at 1:10,000 for 45 min at RT. Localization: Endoplasmic Reticulum cytoplasmic. Staining: KDEL as red fluorescent signal with DAPI blue counterstain. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KDEL Receptor Antibody in Immunocytochemistry (ICC/IF)
          KDEL Receptor Antibody in Western Blot (WB)

          Product Details

          500-301-F86

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, Chicken, Fruit fly, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat, Xenopus

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          KR-10

          Immunogen

          KDEL Receptor Antibody was produced in mice by repeated immunizations raised against a 21 residue synthetic peptide based on the bovine KDEL receptor and the synthetic peptide coupled to KLH.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Storage buffer

          PBS, pH 7.2, with 50% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble endoplasmic reticulum (ER) resident proteins and some membrane proteins. 78 and 94 kDa glucose regulated proteins, GRP78 and GRP94 respectively, and protein disulfide isomerase (PDI) all share the C-terminal KDEL sequence. The presence of carboxy-terminal KDEL appears to be necessary for ER retention and appears to be sufficient to reduce the secretion of proteins from the ER. This retention is reported to be mediated by a KDEL receptor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 108K heat shock protein; 108K HSP; 78 kDa glucose-regulated protein; 78-kD glucose-regulated protein precursor; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250|UniProtKB:P08113}; 98 kDa protein kinase; Binding-immunoglobulin protein; BiP; BiP protein; CG5183-PA; CG5183-PB; CG5183-PC; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; Endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250|UniProtKB:P08113}; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; epididymis secretory sperm binding protein Li 89n; ER lumen protein retaining receptor 1; ER lumen protein retaining receptor 2; ER lumen protein-retaining receptor; ER lumen protein-retaining receptor 1; ERp99; glucose regulated protein, 78 kDa; glucose-regulated protein; glucose-regulated protein GRP94; glucose-regulated protein, 78kDa; gp96 homolog; GRP 78; GRP-78; GRP-94; GRP94; Heat shock 108 kDa protein; heat shock 70 kDa protein 5; heat shock 70 kDa protein 5a; heat shock 70kD protein 5; heat shock 70kD protein 5 (glucose-regulated protein, 78kD); heat shock 70kDa protein 5 (glucose-regulated protein); heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); heat shock protein 108; heat shock protein 108.; heat shock protein 5; Heat shock protein 70 family protein 5; Heat shock protein 90 kDa beta member 1; heat shock protein 90 kDa beta member 1 {ECO:0000250|UniProtKB:P08113}; heat shock protein 90, beta (Grp94), member 1; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; heat shock protein 90kDa beta, member 1; Heat shock protein family A member 5; Heat shock protein family C member 4; heavy-chain binding protein BiP; HSP 108; hsp108; hsp70; HSP70 family protein 5; HSP90 beta; hsp90b1 {ECO:0000250|UniProtKB:P08113}; immunoglobulin binding protein; Immunoglobulin heavy chain-binding protein; KDEL; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 1; KDEL receptor 1; KDEL receptor-like protein; KDEL-receptor; KdelR-PA; KdelR-PB; KdelR-PC; lethal (2) 31Em; Lys-Asp-Glu-Leu; Polymorphic tumor rejection antigen 1; PPK 98; Putative MAPK-activating protein PM23; RP23-446N16.1; Steroidogenesis-activator polypeptide; stress-inducible tumor rejection antigen gp96; Transferrin-binding protein; transforming growth factor alpha regulated gene 2; tumor rejection antigen (gp96) 1; Tumor rejection antigen 1; Tumor rejection antigen gp96; XAP-1 antigen

          View more View less

          Gene Aliases: 8030486F04Rik; AL022860; AU019543; AW215843; baffled; BEST:CK00230; BIP; BiP/Grp78; CG5183; CK00230; D2Wsu141e; D2Wsu17e; Dmel\CG5183; Dmel_CG5183; dmErd2; ECGP; endoplasmin; ERD2; ERD2.1; ERp99; GP96; GRP78; GRP94; HDEL; HEL-S-125m; HEL-S-89n; HEL35; Hsce70; hsp108; HSP90B1; HSPA5; hspa5a; HSPC4; I79_019946; I79_020931; KDELR; KDELR1; l(2)31Em; l(2)k00311; mBiP; MIF2; PM23; ppk98; SEZ-7; Sez7; TA-3; Targ2; Tra-1; TRA1; XAP-1; XELAEV_180384511mg

          View more View less

          UniProt ID: (Pig) Q29092, (Dog) P41148, (Bovine) Q95M18, (Chicken) P08110, (Human) P14625, (Chicken) Q90593, (Bovine) Q0VCX2, (Human) P11021, (Rat) Q66HD0, (Mouse) P08113, (Mouse) P20029, (Chinese hamster) G3I8R9, (Rat) P06761, (Bovine) P33946, (Human) P24390, (Fruit fly) O76767, (Mouse) Q99JH8, (Rat) Q569A6, (Xenopus) Q91883

          View more View less

          Entrez Gene ID: (Pig) 397191, (Rabbit) 100009456, (Dog) 404019, (Bovine) 282646, (Chicken) 374163, (Human) 7184, (Sheep) 101111703, (Chicken) 396487, (Sheep) 780447, (Dog) 480726, (Pig) 407060, (Rabbit) 100008764, (Bovine) 415113, (Human) 3309, (Chinese hamster) 100773331, (Rat) 362862, (Mouse) 22027, (Mouse) 14828, (Chinese hamster) 100689305, (Rat) 25617, (Pig) 100520962, (Bovine) 618184, (Sheep) 101122047, (Human) 10945, (Dog) 484406, (Rabbit) 100349892, (Fruit fly) 34427, (Chinese hamster) 100750912, (Mouse) 68137, (Rat) 361577, (Xenopus) 397850

          View more View less

          Function(s)
          ATP binding RNA binding protein phosphatase binding low-density lipoprotein particle receptor binding unfolded protein binding calcium ion binding protein binding virion binding ATPase activity heat shock protein binding protein binding involved in protein folding misfolded protein binding protein domain specific binding ubiquitin protein ligase binding ribosome binding glycoprotein binding enzyme binding chaperone binding cadherin binding involved in cell-cell adhesion nucleotide binding KDEL sequence binding ER retention sequence binding molecular_function Hsp90 family chaperone membrane trafficking regulatory protein
          Process(es)
          ER-associated ubiquitin-dependent protein catabolic process response to hypoxia retrograde protein transport, ER to cytosol actin rod assembly negative regulation of apoptotic process regulation of phosphoprotein phosphatase activity cellular response to ATP protein folding intracellular sequestering of iron ion toll-like receptor signaling pathway receptor-mediated endocytosis protein transport protein folding in endoplasmic reticulum response to endoplasmic reticulum stress ATF6-mediated unfolded protein response sequestering of calcium ion response to unfolded protein endoplasmic reticulum unfolded protein response negative regulation of protein complex assembly cellular response to unfolded protein protein refolding chaperone mediated protein folding requiring cofactor negative regulation of IRE1-mediated unfolded protein response positive regulation of cell migration posttranslational protein targeting to membrane, translocation maintenance of protein localization in endoplasmic reticulum ER overload response cerebellum structural organization cerebellar Purkinje cell layer development negative regulation of transforming growth factor beta receptor signaling pathway positive regulation of protein ubiquitination cellular response to glucose starvation cellular response to interleukin-4 toxin transport activation of signaling protein activity involved in unfolded protein response substantia nigra development IRE1-mediated unfolded protein response PERK-mediated unfolded protein response regulation of protein folding in endoplasmic reticulum cellular response to antibiotic cellular response to manganese ion negative regulation of protein homodimerization activity cell-cell adhesion regulation of ATF6-mediated unfolded protein response regulation of IRE1-mediated unfolded protein response regulation of PERK-mediated unfolded protein response positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress response to stress positive regulation of embryonic development proteolysis involved in cellular protein catabolic process T cell cytokine production retrograde vesicle-mediated transport, Golgi to ER T cell differentiation T cell apoptotic process protein retention in ER lumen intracellular protein transport ER to Golgi vesicle-mediated transport transport biological_process vesicle-mediated transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0