Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Belongs to the UPF0378 family. 4 isoforms of the human protein are produced by alternative splicing. Song J., Biochim. Biophys. Acta 1760:62-69(2006). Nagase T., DNA Res. 2:37-43(1995). Chen G., Leuk. Res. 29:503-509(2005).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antigen MLAA-22; breast cancer overexpressed gene 1; Breast cancer-overexpressed gene 1 protein; Bridge-like lipid transfer protein family member 2; cancer/testis antigen 101; K0100; Protein hobbit homolog; U937-associated antigen
Gene Aliases: BCOX; BCOX1; BLTP2; CT101; FMP27; KIAA0100
UniProt ID: (Human) Q14667
Entrez Gene ID: (Human) 9703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support