Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
K-RAS (GTPase KRas) belongs to the Ras oncogene family. Ras proteins bind GDP/GTP and possess intrinsic GTPase activity. It plays an important role in the regulation of cell proliferation and has a role in promoting oncogenic events by inducing transcriptional silencing of tumor suppressor genes in colorectal cancer cells in a ZNF304-dependent manner. Mutations in the gene can result in leukemia acute myelogenous, leukemia juvenile myelomonocytic, Noonan Syndrome, gastric cancer, and cardiofaciocutaneous syndrome 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-K-ras; c-Ki-ras; c-Kirsten-ras protein; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; GTPase KRas; K RAS; K-Ras 2; K-ras p21 protein; Ki-Ras; Kirsten rat sarcoma viral oncogene homolog; oncogene KRAS2; PR310 c-K-ras oncogene; RASK; transforming protein p21; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog
Gene Aliases: C-K-RAS; CFC2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; KRAS; KRAS1; KRAS2; NS; NS3; RALD; RASK2
UniProt ID: (Human) P01116
Entrez Gene ID: (Human) 3845
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support