Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: WHANSESSQC NSAELTSPIN MSYNCCSGNF SSRSCGGYLH YPASSCGFSY PSNQVYSTDL CSPSTCQLGS SLYRGCQQTC WEPTSCQTSY VESSPCQTS
In the hair cortex, hair keratin intermediate filaments are embedded in an interfilamentous matrix, consisting of hair keratin-associated proteins (KRTAP), which are essential for the formation of a rigid and resistant hair shaft through their extensive disulfide bond cross-linking with abundant cysteine residues of hair keratins. The matrix proteins include the high-sulfur and high-glycine-tyrosine keratins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: high sulfur keratin associated protein 13.1; High sulfur keratin-associated protein 13.1; Keratin-associated protein 13-1
Gene Aliases: KAP13.1; KRTAP13-1; KRTAP13.1
UniProt ID: (Human) Q8IUC0
Entrez Gene ID: (Human) 140258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support