Search Thermo Fisher Scientific
FIGURE: 1 / 1
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Directed against human herpesvirus 8 (KSHV ORF57) protein. A BLAST analysis was used to suggest cross-reactivity with ORF57 protein from human herpesvirus 8 types P and M sources based on 94% homology with the immunizing sequence. Reactivity against homologues from other sources is unknown.
ORF57 (also known as MTA), one of the earliest Kaposi's sarcoma-associated herpesvirus (KSHV) regulatory proteins to be expressed, is essential for virus lytic replication. A counterpart is present in every herpesvirus sequenced, indicating the importance of this signature viral protein, and those examined act post-transcriptionally, affecting RNA splicing and transport. KSHV ORF57 is capable of establishing both lytic and latent replication cycles. In KS, the virus localizes to tumor progenitor endothelial cells, most of which are latently infected. In cell culture, KSHV replication is generally studied using B-cell lines, such as BCBL-1, generated from primary effusion lymphoma material. Most BCBL-1 cells are latently infected, although there is some spontaneous virus reactivation. Addition of chemical inducers such as sodium n-butyrate, 12-O-tetradecanoylphorbol-13-acetate (TPA), and valproic acid (VA) to these cells efficiently induces the lytic cycle and produces virions. KSHV ORF57 protein predominantly localizes to the nucleus and can shuttle between the nucleus and cytoplasm. Most HSV-1 genes are unspliced; by contrast, ORF57 is spliced gene; the protein is 455 amino acids in length and 50kDa in size.