Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 3T3/A31 RIPA lysate.
DYRK3 belongs to the DYRK family of dual-specificity protein kinases that catalyze phosphorylation of serine/threonine and tyrosine residues. The members of this family share structural similarity, but differ in their substrate specificity, suggesting their involvement in different cellular functions. DYRK3 is a negative regulator of EPO-dependent erythropoiesis, may place an upper limit on red cell production during stress erythropoiesis. Inhibits cell death due to cytokine withdrawal in hematopoietic progenitor cells. May act by regulating CREB/CRE signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dual specificity tyrosine-(Y)-phosphorylation regulated kinase 3; Dual specificity tyrosine-phosphorylation-regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 5; dyrk3; dyrk3 {ECO:0000312|EMBL:AAH97474.1}; KSR; protein kinase Dyrk3; REDK; Regulatory erythroid kinase; RP11-343H5.2; RSU2
Gene Aliases: BC006704; DYRK3; DYRK5; hYAK3-2; RED; REDK
UniProt ID: (Human) O43781, (Rat) Q4V8A3, (Mouse) Q922Y0
Entrez Gene ID: (Human) 8444, (Rat) 304775, (Mouse) 226419
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support