Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Suggested positive control: HEK293 cell lysate.
In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ41162; FLJ42249; FLJ43491; Krc2 1; MGC129855; NGK2; Potassium channel gene 1 (alternative splicing product described in Luneau et al 1991); potassium channel, voltage gated Shaw related subfamily C, member 1; potassium voltage gated channel, Shaw-related subfamily, member 1; Potassium voltage-gated channel subfamily C member 1; potassium voltage-gated channel, Shaw-related subfamily, member 1; RAW2; Shaw-related voltage-gated potassium channel protein 1; voltage-gated potassium channel KCNC1; voltage-gated potassium channel protein KV3.1; Voltage-gated potassium channel subunit Kv3.1; Voltage-gated potassium channel subunit Kv4
Gene Aliases: KCNC1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support