Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Aliquoting is unnecessary for -20°C storage.
LPCAT3 (Lysophosphatidylcholine Acyltransferase 3) is a Protein Coding gene. Diseases associated with LPCAT3 include Retinitis Pigmentosa 40 and Fanconi Anemia, Complementation Group D2. Among its related pathways are triacylglycerol biosynthesis and Glycerophospholipid biosynthesis. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring acyl groups and 1-acylglycerophosphocholine O-acyltransferase activity. An important paralog of this gene is MBOAT1
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphoethanolamine O-acyltransferase; 1-acylglycerophosphoserine O-acyltransferase; LPC acyltransferase 3; lyso-PC acyltransferase 3; lyso-PE acyltransferase; lyso-PS acyltransferase; Lysophosphatidylcholine Acyltransferase 3; lysophosphatidylethanolamine acyltransferase; lysophosphatidylserine acyltransferase; lysophospholipid acyltransferase 12; membrane bound O-acyltransferase domain containing 5; membrane-bound O-acyltransferase domain-containing protein 5; O-acyltransferase (membrane bound) domain containing 5; O-acyltransferase domain-containing protein 5; putative protein similar to nessy; unnamed protein product
Gene Aliases: C3F; Grcc3f; LPCAT; Lpeat; LPLAT 5; LPLAT12; Lplat5; LPSAT; MBOAT5; Moact5; nessy; OACT5; PTG
UniProt ID: (Human) Q6P1A2, (Mouse) Q91V01
Entrez Gene ID: (Pig) 100627653, (Human) 10162, (Mouse) 14792
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support