Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RDKRLEKMV SGCQTRSILE YLRVGGRGGG KGKGRAEGSE KEESRRKRRE RKQRREGGDG EEQDVGDAGR LLLRVLHVSE NPVPLTVRVS PEVRDVRPYI VGAVVRGMDL QPGNALKRFL TSQTKLHEDL CEKRTAATLA THELRAVKGP LLYCARPPQD LKIVPLGRKE AKAKELVRQL QLEAEEQRKQ KKRQSVSGLH RYLHLLDGNE NYPCLVDADG DVISFPPITN SEKTKVKKTT SDLFLEVTSA TSLQICKDVM DALILKMAEM KKYTLENKEE GSLSDTEADA VSGQLPDPTT NPSAGKDGPS LLVVEQVRVV DLEGSLKVVY PSKADLATAP PHVTVVR
LRRC47 gene ontology annotations related to this gene include phenylalanine-tRNA ligase activity; RNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine-rich repeat-containing protein 47; RP1-286D6.3
Gene Aliases: KIAA1185; LRRC47
UniProt ID: (Human) Q8N1G4
Entrez Gene ID: (Human) 57470
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support