Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
LSM12 gene ontology annotations related to this gene include RNA metabolic process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein LSM12; protein LSM12 homolog
Gene Aliases: 1110032E16Rik; 1110059P07; 2600001B17Rik; LSM12; PNAS-135
UniProt ID: (Human) Q3MHD2, (Mouse) Q9D0R8
Entrez Gene ID: (Human) 124801, (Mouse) 268490
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support