Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:100-1:200 | - |
Immunocytochemistry (ICC/IF) |
1:100-1:200 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG2a |
Class |
Monoclonal |
Type |
Antibody |
Clone |
OTI9G8 |
Immunogen |
Full length human recombinant protein of human LTA4H produced in HEK293T cell. |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
0.91 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS with 1% BSA, 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leukotriene A(4) hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA-4 hydrolase; testicular secretory protein Li 27; Tripeptide aminopeptidase LTA4H
Gene Aliases: LTA4; LTA4H
UniProt ID: (Human) P09960
Entrez Gene ID: (Human) 4048
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support