Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-20232 detects leukotriene B4 receptor in human samples.
LTB4R (Leukotriene B4 Receptor) is a member of the GPCR family. It is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response. It has been reported in blood, bone marrow, brain, heart, liver, and skeletal muscle. In addition, it has been shown to be highly expressed in leukocytes and lymphoid tissues such as spleen, thymus, and lymph node. ESTs have been isolated from B-cell/lung/testis, breast, colon, eye, kidney, skin, spleen, testis, and tonsil libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chemoattractant receptor-like 1; chemokine receptor-like 1; G protein-coupled receptor 16; G-protein coupled receptor 16; Leukotriene B4 receptor 1; LTB4-R 1; LTB4-R1; P2Y purinoceptor 7; P2Y7; purinergic receptor P2Y, G-protein coupled, 7
Gene Aliases: BLT; BLT1; BLTR; CMKRL1; GPR16; LTB4R; LTB4R1; LTBR1; P2RY7; P2Y7
UniProt ID: (Human) Q15722
Entrez Gene ID: (Human) 1241
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support