Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: GGAADNTLPN ELHCYDVDFQ TWEVVQPSSD SEVGGAEVPE RACASEEVPT LTYEERVGFK KSRDVFGLDF GTTSAKQPTQ PASELPSGRL FHAAAVISDA MYIFGGTVDN NIRSGEMYRF QFSCYPKCTL HEDYGRLWES RQFCDVEFVL GEKEECVQGH VAIVTARSRW LRRKITQARE RLAQKLEQEA APVPREAPGV AAGGARPPLL HVAIREAEAR PFEVLMQFLY TDKIKYPRKG HVEDVL
LZTR1, a member of the BTB-kelch superfamily, was initially described as a putative transcriptional regulator based on weak homology to members of the basic leucine zipper-like family, the encoded protein subsequently has been shown to localize exclusively to the Golgi network where it may help stabilize the Golgi complex. Deletion of this gene may be associated with DiGeorge syndrome, a developmental field defect involving the third and fourth pharyngeal pouches, causing the absence of thymus and parathyroid glands, congenital cardiac abnormalities and facial dysmorphism. LZTR1 is tyrosine phosphorylated and subsequently degraded upon induction of apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine-zipper-like transcriptional regulator 1; LZTR-1; MGC21205
Gene Aliases: 1200003E21Rik; AI591627; AW550890; Lztr1; Tcfl2
UniProt ID: (Mouse) Q9CQ33
Entrez Gene ID: (Mouse) 66863
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support