Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-29128 detects Lactoferrin from human samples.
PA1-29128 has been successfully used in immunoprecipitation and ELISA applications.
Lactoferrin (LF) is an iron binding glycoprotein. LF consists of a single polypeptide chain (approximately 80 kDa) folded into two structurally homologous lobes, each of which can reversibly bind one ferric ion (Fe3+). LF is found in external fluids, including milk and mucosal secretions, and is a prominent component of secondary granuals of neutrophils. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis. LTF acts as a major first line defense against microbial infections, partly due to its ability to efficiently bind to and hence remove Fe3+ from the environment, but studies have also shown that LTF possesses bactericidal properties and proteolytic activity, capable of cleaving arginine-rich sequences within microbial proteins. Three isoforms of LTF have been identified: LTF-alpha, LTF-beta and LTF-gamma and receptors for LTF are expressed in the gastrointestinal tract, by macrophages, lymphocytes, neutrophils and platelets and also by some bacteria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CKRX; epididymis luminal protein 110; Growth-inhibiting protein 12; Kaliocin 1; Kaliocin-1; Kaliocin1; lactoferricin; Lactoferricin H; Lactoferricin-H; Lactoferrin; lactoferroxin; Lactoferroxin-A; Lactoferroxin-B; Lactoferroxin-C; LactoferroxinA; LactoferroxinB; LactoferroxinC; Lactotransferrin; Lfcin H; Lfcin-H; LfcinH; neutrophil lactoferrin; Talalactoferrin
Gene Aliases: GIG12; LF; LTF
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support