Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 1
Immunogen sequence: EELRSQHDEQ VRLYKLELEQ TYQAKLDSAK LSSDQNDKAA SAAREELKEA RMRLESLSYQ LSGLQKQASA AEDRIRELEE AMAGERDKFR KMLDAKEQEM TEMRDVMQQQ LAEYQELLDV KLALDMEINA YRKLLEGEEE RLKLSPSPSS RVTVSRATSS SSGSLSATGR LGRSKRKRLE VEEPLGSGPS VLGTGTGGSG GFHLAQQASA SGSVSIEEID LEGKFVQLKN NSDKDQSLGN WRIKRQVLEG EEIAYKFTPK YILRAGQMVT VWAAGAGVAH SPPSTLVWKG QSSWGTGESF RTVLVNADGE EVAMRTVKKS SVMRENENGE EEEEEAEFGE EDLFHQQGDP RTTSRGCYVM
Nuclear lamins form a network of intermediate-type filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished, i.e. A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2. Recent evidence has revealed that mutations in A-type lamins give rise to a range of rare but dominant genetic disorders, including Emery-Dreifuss muscular dystrophy, dilated cardiomyopathy with conduction-system disease and Dunnigan-type familial partial lipodystrophy. In addition, the expression of A-type lamins coincides with cell differentiation and as A-type lamins specifically interact with chromatin, a role in the regulation of differential gene expression has been suggested for A-type lamins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lamin B3; Lamin-B2; MGC2721
Gene Aliases: EPM9; LAMB2; LMN2; LMNB2
UniProt ID: (Human) Q03252
Entrez Gene ID: (Human) 84823
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support