Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is pan-specific and reacts well with all Laminin isoforms tested: Laminin-1 (alpha-1, beta-1, and gamma-1) and Laminin-2 (alpha-2, beta-1, and gamma-1).
The antibody functionally inhibits Laminin in mouse and rat. It binds to Laminin and inhibits most, if not all, of its cell adhesion and growth promotive properties. Immunostaining is enhanced by antigen retrieval with pepsin, especially paraffin tissue. Suggested positive control: basement membranes (blood vessels of any tissue, tumors, nerve).
Laminin is a crucial glycoprotein that plays a significant role in the extracellular matrix, particularly within the basement membrane. It is composed of three subunits: alpha, beta, and gamma chains. These chains are disulfide-linked polypeptides that together form a heterotrimer, resembling the shape of a cross. The human genome encodes 11 genetically distinct laminin chains, which combine to form over 14 different laminin isoforms, each with specific tissue distributions and functions. Laminins can bind to other laminin molecules, as well as integrins and other matrix proteins, providing structural support and influencing cell differentiation, migration, and adhesion. These glycoproteins are vital during early embryonic development, where they contribute to the formation of basement membrane-like structures, even in the absence of type IV collagen. The evolutionary origin of laminin gene families can be traced back to early branching animals, providing insights into metazoan gene family development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dystrophia muscularis; Laminin A chain; laminin alpha 2 subunit; laminin B1; Laminin B1 chain; laminin B1 subunit 1; Laminin B2 chain; laminin B2 precursor; Laminin M chain; Laminin subunit alpha-1; Laminin subunit alpha-2; Laminin subunit beta-1; Laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, gamma 1 (formerly LAMB2); laminin-1; Laminin-1 subunit alpha; Laminin-1 subunit beta; Laminin-1 subunit gamma; Laminin-10 subunit beta; Laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit alpha; Laminin-12 subunit beta; Laminin-2 subunit alpha; Laminin-2 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit alpha; Laminin-3 subunit gamma; Laminin-4 subunit alpha; Laminin-4 subunit gamma; Laminin-6 subunit beta; Laminin-6 subunit gamma; Laminin-7 subunit gamma; Laminin-8 subunit beta; Laminin-8 subunit gamma; Laminin-9 subunit gamma; Merosin heavy chain; mutant laminin subunit alpha 2; S-LAM alpha; S-LAM gamma; S-laminin subunit alpha; S-laminin subunit gamma; unnamed protein product
Gene Aliases: 5830440B04; AA408497; B2e; C77966; C80098; C81607; CLM; D130003D08Rik; dy; I79_015129; I79_017505; LAMA; Lama-1; LAMA1; LAMA2; Lamb-1; LAMB1; Lamb1-1; LAMB2; LAMC1; LAMM; LIS5; MDC1A; mer; merosin; mKIAA4087; PTBHS; S-LAM-alpha
UniProt ID: (Human) P25391, (Human) P24043, (Human) P07942, (Human) P11047, (Mouse) P19137, (Mouse) Q60675, (Mouse) P02469
Entrez Gene ID: (Human) 284217, (Sheep) 101102692, (Rabbit) 100357482, (Rabbit) 100350500, (Human) 3908, (Sheep) 101117424, (Human) 3912, (Sheep) 101105743, (Rabbit) 100358679, (Rabbit) 103346447, (Sheep) 101105588, (Human) 3915, (Chinese hamster) 100765555, (Mouse) 16772, (Rat) 316758, (Rat) 309368, (Mouse) 16773, (Mouse) 16777, (Rat) 298941, (Chinese hamster) 100757343, (Rat) 117036, (Chinese hamster) 100761564, (Mouse) 226519
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support