Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
MA5-14713 targets Luteinizing Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples.
The MA5-14713 immunogen is human Luteinizing Hormone.
MA5-14713 detects Luteinizing Hormone which has a predicted molecular weight of approximately 13 kDa.
Product MA514713 is a smaller package size of MIL0102 (formerly sold as a Seradyn product).
LH is a member of the glycoprotein hormone family, Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. LH is expressed in the pituitary gland and promotes spermatogenesis and ovulation by stimulating the testes and ovaries to synthesize steroids. The genes for the beta chains of chorionic gonadotropin and for luteinizing hormone are contiguous on chromosome 19q13.3. Mutations in this gene are associated with hypogonadism which is characterized by infertility and pseudohermaphroditism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anterior pituitary glycoprotein hormones common subunit alpha; CG-alpha; Choriogonadotropin alpha chain; Chorionic gonadotrophin subunit alpha; chorionic gonadotropin, alpha polypeptide; Follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin; Follitropin alpha chain; FSH-alpha; FSH-beta; Glycoprotein hormones alpha chain; hLHB; interstitial cell stimulating hormone, beta chain; LH-B; LSH-alpha; Luteinizing hormone alpha chain; luteinizing hormone beta subunit; Luteinizing hormone subunit beta; Lutropin alpha chain; Lutropin beta chain; Lutropin subunit beta; Thyroid-stimulating hormone alpha chain; Thyrotropin alpha chain; TSH-alpha
Gene Aliases: CG-ALPHA; CGA; CGB4; FSHA; GPHa; GPHA1; HCG; HH23; LHA; LHB; LSH-B; LSH-beta; TSHA
UniProt ID: (Human) P01215, (Human) P01229
Entrez Gene ID: (Human) 1081, (Human) 3972
Molecular Function: peptide hormone intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support