Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Ly-6G/Ly-6C Antibodies

          Invitrogen

          Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), Biotin, eBioscience™

          Advanced Verification
          96 Published Figures
          239 References
          View all (65) Ly-6G/Ly-6C antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), Biotin, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (96)
          • Advanced Verification (1)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 98

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Ly-6G/Ly-6C Antibody (13-5931-82) in Flow

          Staining of mouse bone marrow with Anti-Mouse Ly-6G (Gr-1) FITC and Anti-Human/Mouse CD45R (B220) PE (Product # 12-0452-82). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry (ICC/IF)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry (ICC/IF)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry (ICC/IF)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Ly-6G/Ly-6C Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Immunohistochemistry (IHC)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6G/Ly-6C Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          13-5931-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 24 publications 24 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 7 publications 7 publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 186 publications 186 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          -
          View 3 publications 3 publications

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication

          Affinity Purification (AP)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 12 publications 12 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Dog, Human, Mouse

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), Biotin, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          RB6-8C5

          Conjugate

          Biotin Biotin Biotin
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 561
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • eFluor 506
          • eFluor 660
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine5
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_466800

          Product Specific Information

          Description: The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25 kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations.

          eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (Product # 9668) be used when looking at Ly-6G specific targets.

          Applications Reported: The RB6-8C5 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: The RB6-8C5 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Granulocyte Marker; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN; unnamed protein product

          View more View less

          Gene Aliases: Gr-1; Gr1; Ly-6C; Ly-6C.2; Ly-6C1; Ly-6C2; Ly-6G; Ly6c

          View more View less

          UniProt ID: (Mouse) P0CW02, (Mouse) P0CW03, (Mouse) P35461

          View more View less

          Entrez Gene ID: (Mouse) 17067, (Mouse) 100041546, (Mouse) 546644

          View more View less

          Function(s)
          acetylcholine receptor inhibitor activity acetylcholine receptor binding
          Process(es)
          acetylcholine receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-t4gnh:80/100.66.77.142:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline