Product References
Functional Specificity of the Members of the Sos Family of Ras-GEF Activators: Novel Role of Sos2 in Control of Epidermal Stem Cell Homeostasis.
Cancers
Baltanás FC,Mucientes-Valdivieso C,Lorenzo-Martín LF,Fernández-Parejo N,García-Navas R,Segrelles C,Calzada N,Fuentes-Mateos R,Paramio JM,Bustelo XR,Santos E
MA1-82041 was used in Immunohistochemistry-immunofluorescence to show GEFs Sos1 and Sos2 play specific mechanistic roles in primary mouse keratinocytes.
Thu Apr 29 00:00:00 EDT 2021
Induction of innate cytokine responses by respiratory mucosal challenge with R848 in zebrafish, mice, and humans.
The Journal of allergy and clinical immunology
Progatzky F,Jha A,Wane M,Thwaites RS,Makris S,Shattock RJ,Johansson C,Openshaw PJ,Bugeon L,Hansel TT,Dallman MJ
MA1-82041 was used in Immunohistochemistry-immunofluorescence to demonstrate that respiratory challenge with R848 might offer a novel mucosal model of antiviral immunity.
Mon Jul 01 00:00:00 EDT 2019
Human 3D Gastrointestinal Microtissue Barrier Function As a Predictor of Drug-Induced Diarrhea.
Toxicological sciences : an official journal of the Society of Toxicology
Peters MF,Landry T,Pin C,Maratea K,Dick C,Wagoner MP,Choy AL,Barthlow H,Snow D,Stevens Z,Armento A,Scott CW,Ayehunie S
MA1-82041 was used in Immunohistochemistry to show drug-induced diarrhea can be predicted by analyzing human 3D gastrointestinal microtissue barrier.
Fri Mar 01 00:00:00 EST 2019
Caspase-11 signaling enhances graft-versus-host disease.
Nature communications
Lu Y,Meng R,Wang X,Xu Y,Tang Y,Wu J,Xue Q,Yu S,Duan M,Shan D,Wang Q,Wang H,Billiar TR,Xiao X,Chen F,Lu B
MA1-82041 was used in Proximity Ligation Assay (PLA) to discover how interference with caspase-11 signalling may reduce acute graft-versus-host disease while preserving graft-versus-leukemia activity.
Fri Sep 06 00:00:00 EDT 2019
Spatially Resolved and Quantitative Analysis of VISTA/PD-1H as a Novel Immunotherapy Target in Human Non-Small Cell Lung Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research
Villarroel-Espindola F,Yu X,Datar I,Mani N,Sanmamed M,Velcheti V,Syrigos K,Toki M,Zhao H,Chen L,Herbst RS,Schalper KA
MA1-82041 was used in Immunohistochemistry to support the immunomodulatory role of VISTA in human NSCLC and suggests its potential as therapeutic target.
Sun Apr 01 00:00:00 EDT 2018
Inhibition of TGF-β signaling supports high proliferative potential of diverse p63+ mouse epithelial progenitor cells in vitro.
Scientific reports
Suzuki D,Pinto F,Senoo M
MA1-82041 was used in Immunocytochemistry to show that pharmacological inhibition of TGF-β signalling enables long-term expansion of p63+ epithelial progenitor cells in low Ca2+ media without the need of progenitor cell-purification steps or support by a feeder cell layer.
Thu Jul 20 00:00:00 EDT 2017
Label-free isolation of prostate circulating tumor cells using Vortex microfluidic technology.
NPJ precision oncology
Renier C,Pao E,Che J,Liu HE,Lemaire CA,Matsumoto M,Triboulet M,Srivinas S,Jeffrey SS,Rettig M,Kulkarni RP,Di Carlo D,Sollier-Christen E
MA1-82041 was used in Immunocytochemistry to describe the use of the microfluidic Vortex Chip for size-based isolation of circulating tumor cells (CTCs) from 22 patients with advanced prostate cancer and, from an enumeration study on 18 of these patients, show that we can capture CTCs with high purity and efficiency in less than 1 h.
Thu Oct 01 00:00:00 EDT 2020
Human breast cancer biopsies induce eosinophil recruitment and enhance adjacent cancer cell proliferation.
Breast cancer research and treatment
Szalayova G,Ogrodnik A,Spencer B,Wade J,Bunn J,Ambaye A,James T,Rincon M
MA182041 was used in immunohistochemistry - paraffin section to investigate inflammatory responses present at the breast cancer biopsy wound site
Wed Jun 01 00:00:00 EDT 2016
Differentiation of equine induced pluripotent stem cells into a keratinocyte lineage.
Equine veterinary journal
Aguiar C,Therrien J,Lemire P,Segura M,Smith LC,Theoret CL
MA1-80241 was used in immunocytochemistry to investigate the possibility eiPSC differentiation into equine keratinocytes (eiPSC-KC), a stem cell application with potential skin regeneration implications.
Sun May 01 00:00:00 EDT 2016
A Previously Undescribed Presentation of Mixed Adenoneuroendocrine Carcinoma.
Case reports in pathology
De Luca-Johnson J,Zenali M
MA182041 was used in immunohistochemistry to discuss a case of mixed adenoneuroendocrine carcinoma of stomach with tubular adenoma and well-differentiated neuroendocrine tumor in the primary tumor in the stomach
Fri Apr 08 00:00:00 EDT 2022
The first microbial environment of infants born by C-section: the operating room microbes.
Microbiome
Shin H,Pei Z,Martinez KA,Rivera-Vinas JI,Mendez K,Cavallin H,Dominguez-Bello MG
MA1-82041 was used in immunohistochemistry - paraffin section to test if the operating room environment contains human skin bacteria that could be seeding C-section born infants
Tue Dec 01 00:00:00 EST 2015
The carboxy-terminus of p63 links cell cycle control and the proliferative potential of epidermal progenitor cells.
Development (Cambridge, England)
Suzuki D,Sahu R,Leu NA,Senoo M
MA1-82041 was used in immunohistochemistry - paraffin section to determine the link between cell cycle control and proliferative potential of epidermal progenitor cells by the carboxy-terminus of p63
Thu Jan 15 00:00:00 EST 2015
Investigation of the origin of stromal and endothelial cells at the desmoplastic interface in xenograft tumor in mice.
Pathology, research and practice
Jung M,Ryu YJ,Kang G
MA1-82041 was used in immunohistochemistry - paraffin section to determine the desmoplastic interface in xenograft tumor in mice comprised of stromal and endothelial cells
Tue Dec 01 00:00:00 EST 2015
Establishment and characterization of a continuous cell line from thymus of striped snakehead, Channa striatus (Bloch 1793).
In vitro cellular & developmental biology. Animal
Sood N,Chaudhary DK,Pradhan PK,Verma DK,Raja Swaminathan T,Kushwaha B,Punia P,Jena JK
MA1-82041 was used in immunocytochemistry to generate and characterize a thymic cell line derived from Channa striatus.
Tue Sep 01 00:00:00 EDT 2015
miR-634 restores drug sensitivity in resistant ovarian cancer cells by targeting the Ras-MAPK pathway.
Molecular cancer
van Jaarsveld MT,van Kuijk PF,Boersma AW,Helleman J,van IJcken WF,Mathijssen RH,Pothof J,Berns EM,Verweij J,Wiemer EA
MA1-82041 was used in immunohistochemistry to determine how sensitivity is restored in resistant ovarian cancer cells by targeting the Ras-MAPK pathway by miR-634
Tue Nov 17 00:00:00 EST 2015
Immunohistochemical expression of cytokeratins and epithelial membrane protein 2 in nasopharyngeal carcinoma and its potential implications.
Asian Pacific journal of cancer prevention : APJCP
Ahmed HG,Abdul Gader Suliman RS,Abd El Aziz MS,Alshammari FD
MA1-82041 was used in immunohistochemistry - paraffin section to identify biomarkers for nasopharyngeal carcinoma
Mon Dec 14 00:00:00 EST 2015
Increased utilization, verification, and clinical implications of immunocytochemistry: Experience in a northern New England hospital.
Diagnostic cytopathology
Sauter JL,Ambaye AB,Mount SL
MA1-82041 was used in immunohistochemistry to study a northern New England hospital and increased utilization, verification, and clinical implications of immunocytochemistry
Tue Sep 01 00:00:00 EDT 2015
Fumarate induces redox-dependent senescence by modifying glutathione metabolism.
Nature communications
Zheng L,Cardaci S,Jerby L,MacKenzie ED,Sciacovelli M,Johnson TI,Gaude E,King A,Leach JD,Edrada-Ebel R,Hedley A,Morrice NA,Kalna G,Blyth K,Ruppin E,Frezza C,Gottlieb E
MA1-82041 was used in immunohistochemistry - paraffin section to study how redox-dependent senescence is induced by fumarate to modify glutathione metabolism
Fri Jan 23 00:00:00 EST 2015
Keratinocyte growth factor-2 and autologous serum potentiate the regenerative effect of mesenchymal stem cells in cornea damage in rats.
International journal of ophthalmology
Pınarlı FA,Okten G,Beden U,Fışgın T,Kefeli M,Kara N,Duru F,Tomak L
MA1-82041 was used in immunohistochemistry to study the use of mesenchymal stem cells to repair rat corneal damage and the additional therapeutic effect obtained from the use of KGF-2 and autologous serum
Mon May 05 00:00:00 EDT 2014
Invasive ductular carcinoma in 2 rhesus macaques (Macaca mulatta).
Comparative medicine
Beck AP,Brooks A,Zeiss CJ
MA1-82041 was used in immunohistochemistry - paraffin section to assess the prognostic value of immunohistochemical markers in nonhuman primates
Fri Aug 01 00:00:00 EDT 2014
Superficial soft tissue biphasic synovial sarcoma with apocrine differentiation in the glandular component: a report of two cases.
The American Journal of dermatopathology
Shelekhova KV,Calonje E,Grossmann P,Kacerovska D,Koudela K,Mirka H,Michal M,Kazakov DV
MA1-82041 was used in immunohistochemistry to report on two cases of superficial soft tissue biphasic synovial sarcoma presenting with apocrine differentiation
Wed Oct 01 00:00:00 EDT 2014
Dietary cholesterol directly induces acute inflammasome-dependent intestinal inflammation.
Nature communications
Progatzky F,Sangha NJ,Yoshida N,McBrien M,Cheung J,Shia A,Scott J,Marchesi JR,Lamb JR,Bugeon L,Dallman MJ
Published figure using Cytokeratin Pan Type I/II cocktail antibody (Product # MA1-82041) in Flow Cytometry
Tue Dec 23 00:00:00 EST 2014
Liquid-based cytologic findings of solitary extramedullary plasmacytoma in thyroid: a case report identified with fine-needle aspiration cytology.
Diagnostic cytopathology
Lee CH,Jung YY,Chung YR,Ryu HS
MA1-82041 was used in immunocytochemistry to report on a case of solitary extramedullary plasmacytoma of the thyroid
Sat Nov 01 00:00:00 EDT 2014
Poor-prognosis colon cancer is defined by a molecularly distinct subtype and develops from serrated precursor lesions.
Nature medicine
De Sousa E Melo F,Wang X,Jansen M,Fessler E,Trinh A,de Rooij LP,de Jong JH,de Boer OJ,van Leersum R,Bijlsma MF,Rodermond H,van der Heijden M,van Noesel CJ,Tuynman JB,Dekker E,Markowetz F,Medema JP,Vermeulen L
MA1-82041 was used in immunohistochemistry to study a colon cancer subtype related to sessile-serrated adenomas and which has a poor prognosis
Wed May 01 00:00:00 EDT 2013
Efficient generation of human iPSCs by a synthetic self-replicative RNA.
Cell stem cell
Yoshioka N,Gros E,Li HR,Kumar S,Deacon DC,Maron C,Muotri AR,Chi NC,Fu XD,Yu BD,Dowdy SF
MA1-82041 was used in immunohistochemistry to develop a synthetic self-replicating RNA replicon expressing reprogramming factors that enables the efficient generation of induced pluripotent stem cells
Thu Aug 01 00:00:00 EDT 2013
Epidermal adrenergic signaling contributes to inflammation and pain sensitization in a rat model of complex regional pain syndrome.
Pain
Li W,Shi X,Wang L,Guo T,Wei T,Cheng K,Rice KC,Kingery WS,Clark JD
MA1-82041 was used in immunohistochemistry to use a rat model to study inflammation and sensitization to pain in complex regional pain syndrome and the role played by adrenergic signaling
Thu Aug 01 00:00:00 EDT 2013
T-Cell tropism of simian varicella virus during primary infection.
PLoS pathogens
Ouwendijk WJ,Mahalingam R,de Swart RL,Haagmans BL,van Amerongen G,Getu S,Gilden D,Osterhaus AD,Verjans GM
MA1-82041 was used in immunohistochemistry to study the role of memory T-cells in the spread of SVV to target organs during primary infection
Wed May 01 00:00:00 EDT 2013
Epstein-Barr virus transcytosis through polarized oral epithelial cells.
Journal of virology
Tugizov SM,Herrera R,Palefsky JM
MA1-82041 was used in immunocytochemistry to study the mechanism by which EBV transmigrates through oral epithelial cells
Mon Jul 01 00:00:00 EDT 2013
Malignant transformation of adenomyoepithelioma of the breast by a monophasic population: a report of two cases and review of literature.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica
Marian C,Boila A,Soanca D,Malau M,Podeanu DM,Resetkova E,Stolnicu S
MA1-82041 was used in immunohistochemistry to report on two cases of malignant transformation of adenomyoepithelioma of the breast by a monophasic population
Mon Apr 01 00:00:00 EDT 2013
Dural angioleiomyoma of the middle cranial fossa: a case report and review of the literature.
Brain tumor pathology
Zhou Z,Yu M,Yang S,Zhou J,Sun R,Yang G
MA1-82041 was used in immunohistochemistry to report on a case of dural angioleiomyoma of the middle cranial fossa
Mon Apr 01 00:00:00 EDT 2013
The effects of hyperbaric oxygen application against cholestatic oxidative stress and hepatic damage after bile duct ligation in rats.
The Journal of surgical research
Ayvaz S,Kanter M,Aksu B,Sahin SH,Uzun H,Erboga M,Pul M
MA1-82041 was used in immunohistochemistry to study the mechanisms underlying the beneficial effects of hyperbaric oxygen against oxidative and cholestatic hepatic damage caused by bile duct ligation in rats
Mon Jul 01 00:00:00 EDT 2013
Malignant ossifying fibromyxoid tumor of the tongue: case report and review of the literature.
Head & face medicine
Ohta K,Taki M,Ogawa I,Ono S,Mizuta K,Fujimoto S,Takata T,Kamata N
MA1-82041 was used in immunohistochemistry to characterize a case of malignant ossifying fibromyxoid tumor in the tongue
Mon Jun 24 00:00:00 EDT 2013
Invasive thymoma metastatic to the cavernous sinus.
Surgical neurology international
Nassiri F,Scheithauer BW,Corwin DJ,Kaplan HG,Mayberg M,Cusimano MD,Rotondo F,Kovacs K
MA1-82041 was used in immunohistochemistry to present the third case of thymoma and the second invasive thymoma to metastasize to the cavernous sinus and adjacent to the pituitary
Wed Jun 19 00:00:00 EDT 2013
Is mammary not otherwise specified-type sarcoma with CD10 expression a distinct entity? A rare case report with immunohistochemical and ultrastructural study.
Diagnostic pathology
Yang GZ,Li J,Jin H,Ding HY
MA1-82041 was used in immunohistochemistry to present a case of not otherwise specified-type sarcoma with CD10 expression in the left breast.
Mon Jan 28 00:00:00 EST 2013
Diagnostic and therapeutic strategy and the most efficient prognostic factors of breast malignant fibrous histiocytoma.
Scientific reports
Qiu SQ,Wei XL,Huang WH,Wu MY,Qin YS,Li YK,Zhang GJ
MA1-82041 was used in immunohistochemistry - paraffin section to report the clinicopathological features of 9 breast malignant fibrous histiocytoma patients
Fri Feb 14 00:00:00 EST 2014
Sentinel nodes identified by computed tomography-lymphography accurately stage the axilla in patients with breast cancer.
BMC medical imaging
Motomura K,Sumino H,Noguchi A,Horinouchi T,Nakanishi K
MA1-82041 was used in immunohistochemistry to test if computed tomography-lymphography distinguishes sentinel nodes from non-sentinel nodes in patients with breast cancer
Mon Dec 09 00:00:00 EST 2013
Infectious salmon anaemia virus infection of Atlantic salmon gill epithelial cells.
Virology journal
Weli SC,Aamelfot M,Dale OB,Koppang EO,Falk K
MA1-82041 was used in immunocytochemistry to study cell tropism of infectious salmon anemia virus
Wed Jan 02 00:00:00 EST 2013
Correlation between the area of high-signal intensity on SPIO-enhanced MR imaging and the pathologic size of sentinel node metastases in breast cancer patients with positive sentinel nodes.
BMC medical imaging
Motomura K,Izumi T,Tateishi S,Sumino H,Noguchi A,Horinouchi T,Nakanishi K
MA1-82041 was used in immunohistochemistry - paraffin section to determine the predictive criteria of the size of nodal metastases with superparamagnetic iron oxide-enhanced MR imaging in breast cancer
Fri Sep 13 00:00:00 EDT 2013
Giant angiofibromas in tuberous sclerosis complex: a possible role for localized lymphedema in their pathogenesis.
Journal of the American Academy of Dermatology
Kacerovska D,Kerl K,Michal M,Filipova H,Vrtel R,Vanecek T,Zelenakova H,Kraus J,Kodet R,Kazakov DV
MA1-82041 was used in immunohistochemistry to study giant angiofibromas in tuberous sclerosis complex
Sat Dec 01 00:00:00 EST 2012
Smooth muscle tension induces invasive remodeling of the zebrafish intestine.
PLoS biology
Seiler C,Davuluri G,Abrams J,Byfield FJ,Janmey PA,Pack M
MA1-82041 was used in immunohistochemistry to study epithelial cell invasion of the zebrafish intestine in response to smooth muscle tension and the underlying mechanisms
Wed Jun 19 00:00:00 EDT 2013
Substance P signaling controls mast cell activation, degranulation, and nociceptive sensitization in a rat fracture model of complex regional pain syndrome.
Anesthesiology
Li WW,Guo TZ,Liang DY,Sun Y,Kingery WS,Clark JD
MA1-82041 was used in immunohistochemistry to study the role of substance P signaling via the neurokinin-1 receptor in mast cell function and nociception in a model of complex regional pain syndrome
Sun Apr 01 00:00:00 EDT 2012
A novel function for p21Cip1 and acetyltransferase p/CAF as critical transcriptional regulators of TGFβ-mediated breast cancer cell migration and invasion.
Breast cancer research : BCR
Dai M,Al-Odaini AA,Arakelian A,Rabbani SA,Ali S,Lebrun JJ
MA1-82041 was used in immunohistochemistry to study the transcriptional regulation of the TGF-beta-mediated migration and invasion of breast cancer cells by p21Cip1 and acetyltransferase p/CAF
Thu Sep 20 00:00:00 EDT 2012
FTIR microscopy of biological cells and tissue: data analysis using resonant Mie scattering (RMieS) EMSC algorithm.
The Analyst
Bassan P,Sachdeva A,Kohler A,Hughes C,Henderson A,Boyle J,Shanks JH,Brown M,Clarke NW,Gardner P
MA1-82041 was used in immunohistochemistry to develop an algorithm for analyzing the data from FTIR microscopy of cells and tissues
Wed Mar 21 00:00:00 EDT 2012
Low-temperature electrospun silk scaffold for in vitro mucosal modeling.
Journal of biomedical materials research. Part A
Bulysheva AA,Bowlin GL,Klingelhutz AJ,Yeudall WA
MA1-82041 was used in immunocytochemistry to evaluate the application of cryogenic electrospun silk scaffold in an in vitro mucosal model
Thu Mar 01 00:00:00 EST 2012
Localization of estrogen receptor in the central lymphoid organs of chickens during the late stage of embryogenesis.
Bioscience, biotechnology, and biochemistry
Katayama M,Fukuda T,Narabara K,Abe A,Kondo Y
MA1-82041 was used in immunohistochemistry to study estrogen receptor expression in developing chick embryo central lymphoid tissue and the significance for immune function
Fri May 17 00:00:00 EDT 2013
Rgs19 regulates mouse palatal fusion by modulating cell proliferation and apoptosis in the MEE.
Mechanisms of development
Sohn WJ,Ji YR,Kim HS,Gwon GJ,Chae YM,An CH,Park HD,Jung HS,Ryoo ZY,Lee S,Kim JY
MA1-82041 was used in immunohistochemistry to study the mechanism by which Rgs19 regulates mouse palatal fusion
Wed Jun 12 00:00:00 EDT 2013
Validation of three breast cancer nomograms and a new formula for predicting non-sentinel lymph node status.
Asian Pacific journal of cancer prevention : APJCP
Derici S,Sevinc A,Harmancioglu O,Saydam S,Kocdor M,Aksoy S,Egeli T,Canda T,Ellidokuz H,Derici S
MA1-82041 was used in immunohistochemistry to study the ability of three current breast cancer nomograms to predict non-sentinel lymph node status and to compare the results with those of a novel calculation
Sat Apr 23 00:00:00 EDT 2016
Sporadic haemangioblastoma of the kidney with rhabdoid features and focal CD10 expression: report of a case and literature review.
Diagnostic pathology
Yin WH,Li J,Chan JK
MA1-82041 was used in immunohistochemistry to report on a case of sporadic haemangioblastoma of the kidney with rhabdoid features and focal CD10 expression
Thu Apr 12 00:00:00 EDT 2012
Keratinocyte expression of inflammatory mediators plays a crucial role in substance P-induced acute and chronic pain.
Journal of neuroinflammation
Wei T,Guo TZ,Li WW,Hou S,Kingery WS,Clark JD
MA1-82041 was used in immunohistochemistry to study the role of keratinicyte inflammatory mediators in substance P-induced acute and chronic pain
Mon Jul 23 00:00:00 EDT 2012
A novel TP53 somatic mutation involved in the pathogenesis of pediatric choroid plexus carcinoma.
Medical science monitor : international medical journal of experimental and clinical research
Lv SQ,Song YC,Xu JP,Shu HF,Zhou Z,An N,Huang QL,Yang H
MA1-82041 was used in immunohistochemistry - paraffin section to review features of choroid plexus carcinoma
Tue May 01 00:00:00 EDT 2012
Distinct histological features characterize primary angiosarcoma of bone.
Histopathology
Verbeke SL,Bertoni F,Bacchini P,Sciot R,Fletcher CD,Kroon HM,Hogendoorn PC,Bovée JV
MA1-82041 was used in immunohistochemistry to define the histological features of primary angiosarcoma of bone
Sat Jan 01 00:00:00 EST 2011
Congenital stridor in the context of Chiari malformation type II: the etiological role of vernix caseosa granulomatous meningitis.
Journal of neurosurgery. Pediatrics
Stritzke AI,Dunham CP,Smyth JA,Steinbok P
MA1-82041 was used in immunohistochemistry to describe a patient with a low lumbar neural tube defect and features of Chiari malformation type II
Sat Oct 01 00:00:00 EDT 2011
Molecular characterization of an EWSR1-POU5F1 fusion associated with a t(6;22) in an undifferentiated soft tissue sarcoma.
Cancer genetics
Deng FM,Galvan K,de la Roza G,Zhang S,Souid AK,Stein CK
MA1-82041 was used in immunohistochemistry to characterize a clinical case of undifferentiated soft tissue sarcoma with chromosomal rearrangement and resultant EWSR1-POU5F1 fusion
Mon Aug 01 00:00:00 EDT 2011
Morphological evidences in circumvallate papilla and von Ebners' gland development in mice.
Anatomy & cell biology
Sohn WJ,Gwon GJ,An CH,Moon C,Bae YC,Yamamoto H,Lee S,Kim JY
MA1-82041 was used in immunohistochemistry - paraffin section to investigate the positioning of taste buds in circumvallate papilla and branching morphogenesis of von Ebners' gland in tongue development
Thu Dec 01 00:00:00 EST 2011
Histological assessment of PAXgene tissue fixation and stabilization reagents.
PloS one
Kap M,Smedts F,Oosterhuis W,Winther R,Christensen N,Reischauer B,Viertler C,Groelz D,Becker KF,Zatloukal K,Langer R,Slotta-Huspenina J,Bodo K,de Jong B,Oelmuller U,Riegman P
MA1-82041 was used in immunohistochemistry - paraffin section to examine the suitability of PAXgene tissue fixation for (immuno)histological methods
Mon Mar 26 00:00:00 EDT 2012
Intra-uterine tissue engineering of full-thickness skin defects in a fetal sheep model.
Biomaterials
Hosper NA,Eggink AJ,Roelofs LA,Wijnen RM,van Luyn MJ,Bank RA,Harmsen MC,Geutjes PJ,Daamen WF,van Kuppevelt TH,Tiemessen DM,Oosterwijk E,Crevels JJ,Blokx WA,Lotgering FK,van den Berg PP,Feitz WF
MA1-82041 was used in immunohistochemistry to evaluate the efficacy of collagen scaffolds in the intra-uterine treatment
Sat May 01 00:00:00 EDT 2010
Fracture induces keratinocyte activation, proliferation, and expression of pro-nociceptive inflammatory mediators.
Pain
Li WW,Guo TZ,Li XQ,Kingery WS,Clark DJ
MA1-82041 was used in immunohistochemistry to study the cellular and protein expression changes induced by fracture
Wed Dec 01 00:00:00 EST 2010
Exacerbated innate host response to SARS-CoV in aged non-human primates.
PLoS pathogens
Smits SL,de Lang A,van den Brand JM,Leijten LM,van IJcken WF,Eijkemans MJ,van Amerongen G,Kuiken T,Andeweg AC,Osterhaus AD,Haagmans BL
MA1-82041 was used in immunohistochemistry to study the pathogenesis of SARS coronavirus in aged macaques
Fri Feb 05 00:00:00 EST 2010
Ventilator-induced inflammatory response in lipopolysaccharide-exposed rat lung is mediated by angiotensin-converting enzyme.
The American journal of pathology
Wösten-van Asperen RM,Lutter R,Specht PA,van Woensel JB,van der Loos CM,Florquin S,Lachmann B,Bos AP
MA1-82041 was used in immunohistochemistry to investigate the role of angiotensin-converting enzyme in ventilator-induced inflammatory response
Sat May 01 00:00:00 EDT 2010
Antiproliferative and apoptotic effects of the herbal agent Pygeum africanum on cultured prostate stromal cells from patients with benign prostatic hyperplasia (BPH).
The Prostate
Quiles MT,Arbós MA,Fraga A,de Torres IM,Reventós J,Morote J
MA1-82041 was used in immunocytochemistry to investigate the effect of specific herbal agent on prostate cell function from benign prostatic hyperplasia patients
Thu Jul 01 00:00:00 EDT 2010
Induction of invasion in an organotypic oral cancer model by CoCl2, a hypoxia mimetic.
European journal of oral sciences
Brusevold IJ,Husvik C,Schreurs O,Schenck K,Bryne M,Søland TM
MA1-82041 was used in immunohistochemistry to develop an in vitro model for experimental studies of cancer cell invasion
Thu Apr 01 00:00:00 EDT 2010
Post-junctional facilitation of Substance P signaling in a tibia fracture rat model of complex regional pain syndrome type I.
Pain
Wei T,Li WW,Guo TZ,Zhao R,Wang L,Clark DJ,Oaklander AL,Schmelz M,Kingery WS
MA1-82041 was used in immunohistochemistry to investigate the influence of substance P activation on specific phenotypes in injured limb
Sat Aug 01 00:00:00 EDT 2009
The NALP1 inflammasome controls cytokine production and nociception in a rat fracture model of complex regional pain syndrome.
Pain
Li WW,Guo TZ,Liang D,Shi X,Wei T,Kingery WS,Clark JD
MA1-82041 was used in immunohistochemistry to investigate the role of the NALP1 inflammasome in cytokine production and nociception
Tue Dec 15 00:00:00 EST 2009
Deletion of the ectodomain unleashes the transforming, invasive, and tumorigenic potential of the MET oncogene.
Cancer science
Merlin S,Pietronave S,Locarno D,Valente G,Follenzi A,Prat M
MA1-82041 was used in immunohistochemistry to examine the tumorigenic effect of the MET oncogene
Wed Apr 01 00:00:00 EDT 2009
Pulmonary meningothelial-like nodules: new insights into a common but poorly understood entity.
The American journal of surgical pathology
Mukhopadhyay S,El-Zammar OA,Katzenstein AL
MA1-82041 was used in immunohistochemistry to study pulmonary meningothelial-like nodules
Wed Apr 01 00:00:00 EDT 2009
Colony-stimulating factor-1-dependent macrophage functions regulate the maternal decidua immune responses against Listeria monocytogenes infections during early gestation in mice.
Infection and immunity
Qiu X,Zhu L,Pollard JW
MA1-82041 was used in immunohistochemistry to investigate the influence of colony-stimulating factor-1 on macrophage function and L. monocytogenes infection treatment
Thu Jan 01 00:00:00 EST 2009
Primary choroid plexus papilloma of the sacral nerve roots.
Journal of neurosurgery. Spine
Boldorini R,Panzarasa G,Girardi P,Monga G
MA1-82041 was used in immunohistochemistry to report on a case of choroid plexus papilloma of the sacral nerve roots
Thu Jan 01 00:00:00 EST 2009
Rhabdomyosarcoma arising in a mature cystic teratoma with contralateral serous carcinoma: case report and review of the literature.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists
Kefeli M,Kandemir B,Akpolat I,Yildirim A,Kokcu A
MA1-82041 was used in immunohistochemistry to report on a case of rhabdomyosarcoma arising in a mature cystic teratoma with contralateral serous carcinoma
Wed Jul 01 00:00:00 EDT 2009
Metastasis of adenocarcinoma of the gall bladder to an endometrial polyp detected by endometrial curettage: case report and review of the literature.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists
Kefeli M,Gonullu G,Can B,Malatyalioglu E,Kandemir B
MA1-82041 was used in immunohistochemistry to report on a case of metastasis of gall bladder adenocarcinoma to an endometrial polyp
Wed Jul 01 00:00:00 EDT 2009
The clinical significance of tumor cells in bone marrow or apheresis product and the efficacy of CD34+ selection and high-dose chemotherapy in patients with Stage III breast cancer.
Journal of clinical apheresis
Arpaci F,Safali M,Ozet A,Ural AU,Cetin T,Ozturk M,Ataergin S,Kuzhan O,Komurcu S,Ozturk B,Acikel C
MA1-82041 was used in immunohistochemistry to study the presence of tumor cells in bone marrow and its clinical significance in breast cancer treatment
Thu Jan 07 00:00:00 EST 2010
Multifocal metanephric adenoma in childhood.
Pathology international
Kohashi K,Oda Y,Nakamori M,Yamamoto H,Tamiya S,Toubo T,Kinoshita Y,Tajiri T,Taguchi T,Tsuneyoshi M
MA1-82041 was used in immunohistochemistry to report a clinical case of childhood multifocal metanephric adenoma
Thu Jan 01 00:00:00 EST 2009
Carcinoid-like pattern in melanoma: report of 4 cases.
The American Journal of dermatopathology
Kacerovska D,Michal M,Sosna B,Cempirkova D,Ambrus M,Richtr P,Danis D,Zelger B,Kazakov DV
MA1-82041 was used in immunohistochemistry to report on four cases of carcinoid-like pattern in melanoma
Sat Aug 01 00:00:00 EDT 2009
Epidermotropic chondroid metastasis of melanoma: report of a case of metastatic melanoma with previously unreported morphological features.
The American Journal of dermatopathology
Piana S,Valli R,Ricci C
MA1-82041 was used in immunohistochemistry to report a clinical case of metastatic melanoma
Fri May 01 00:00:00 EDT 2009
Giant cell tumor-like lesion of the urinary bladder: a report of two cases and literature review; giant cell tumor or undifferentiated carcinoma?
Diagnostic pathology
Behzatoğlu K,Durak H,Canberk S,Aydin O,Huq GE,Oznur M,Ozyalvaçli G,Yildiz P
MA1-82041 was used in immunohistochemistry to report on two cases of giant cell tumor-like lesion of the urinary bladder
Thu Dec 31 00:00:00 EST 2009
Targeting a single mismatched minor histocompatibility antigen with tumor-restricted expression eradicates human solid tumors.
Blood
Hambach L,Vermeij M,Buser A,Aghai Z,van der Kwast T,Goulmy E
MA1-82041 was used in immunohistochemistry to examine the graft-versus-host disease during allogeneic human leukocyte antigen-matched stem cell transplantation
Mon Sep 01 00:00:00 EDT 2008
Membrane type 1 matrix metalloproteinase-mediated stromal syndecan-1 shedding stimulates breast carcinoma cell proliferation.
Cancer research
Su G,Blaine SA,Qiao D,Friedl A
MA1-82041 was used in immunocytochemistry to investigate the effect of fibroblast-derived membrane type 1 matrix metalloproteinase on stromal-epithelial signaling in breast carcinomas
Sat Nov 15 00:00:00 EST 2008
Epithelial and pseudoepithelial differentiation in glioblastoma and gliosarcoma: a comparative morphologic and molecular genetic study.
Cancer
Rodriguez FJ,Scheithauer BW,Giannini C,Bryant SC,Jenkins RB
MA1-82041 was used in immunohistochemistry - paraffin section to discuss the morphological diversity of glioblastomas
Sat Nov 15 00:00:00 EST 2008
Adenomatoid tumour of the liver.
Journal of clinical pathology
Nagata S,Aishima S,Fukuzawa K,Takagi H,Yonemasu H,Iwashita Y,Kinoshita T,Wakasugi K
MA1-82041 was used in immunohistochemistry to report on a case of adenomatoid tumour of the liver
Sun Jun 01 00:00:00 EDT 2008
The coexistence of invasive ductal carcinoma and malignant phyllodes tumor with liposarcomatous and chondrosarcomatous differentiation in the same breast in a post-osteosarcoma case.
Pathology, research and practice
Kefeli M,Yildiz L,Akpolat I,Balci P,Ozen N
MA1-82041 was used in immunohistochemistry to report on a patient with both invasive ductal carcinoma and malignant phyllodes tumor with liposarcomatous and chondrosarcomatous differentiation
Tue Mar 03 00:00:00 EST 2009
Clinocopathologic and immunohistochemical findings of multiple genital leiomyomas and mammary adenocarcinomas in a bitch.
Reproduction in domestic animals = Zuchthygiene
Ozmen O,Haligur M,Kocamuftuoglu M
MA1-82041 was used in immunohistochemistry to examine a case of canine multiple genital leiomyomas and mammary adenocarcinomas
Sun Jun 01 00:00:00 EDT 2008
Eccrine syringofibroadenoma associated with well-differentiated squamous cell carcinoma.
The American Journal of dermatopathology
Kacerovska D,Nemcova J,Michal M,Kazakov DV
MA1-82041 was used in immunohistochemistry to report on a case of eccrine syringofibroadenoma associated with well-differentiated squamous cell carcinoma
Mon Dec 01 00:00:00 EST 2008
Plasmoblastic lymphoma associated with human immunodeficiency virus.
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie
Horváth E,Krenács L,Bagdi E,Pávai Z,Macarie I,Nagy EE,Demian S
MA1-82041 was used in immunohistochemistry to investigate the association of plasmoblastic lymphoma with human immunodeficiency virus
Thu Dec 11 00:00:00 EST 2008
Chromophobe renal cell carcinoma with osseous metaplasia containing fatty bone marrow element: a case report.
Pathology, research and practice
Kefeli M,Yildiz L,Aydin O,Kandemir B,Faik Yilmaz A
MA1-82041 was used in immunohistochemistry to report on a case of chromophobe renal cell carcinoma with osseous metaplasia containing fatty bone marrow element
Fri Nov 09 00:00:00 EST 2007
Myointimoma of the glans penis.
Pathology international
Vardar E,Gunlusoy B,Arslan M,Kececi S
MA1-82041 was used in immunohistochemistry to report on a case of myointimoma of the glans penis
Thu Mar 01 00:00:00 EST 2007
Atypical reactive ovarian surface epithelium, a pitfall in pathologic assessment.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society
Aydin NE,Celik O,Hasçalik S,Edali MN
MA1-82041 was used in immunohistochemistry to report the clinical features of a patient with atypical reactive ovarian surface epithelium
Wed Jul 26 00:00:00 EDT 2006
Re-evaluation of axillary skip metastases in the era of sentinel lymph node biopsy in breast cancer.
Surgery today
Keskek M,Balas S,Gokoz A,Sayek I
MA1-82041 was used in immunohistochemistry to investigate the axillary skip metastases in breast cancer
Thu Nov 01 00:00:00 EDT 2007
Malignant perivascular epithelioid cell tumor of the colon: report of a case with molecular analysis.
Pathology international
Yamamoto H,Oda Y,Yao T,Oiwa T,Kobayashi C,Tamiya S,Kawaguchi K,Hino O,Tsuneyoshi M
MA1-82041 was used in immunohistochemistry to report on a case of malignant perivascular epitheloid cell tumor of the colon
Sun Jan 01 00:00:00 EST 2006
Identification of major cell types in paraffin sections of bovine tissues.
BMC veterinary research
Niku M,Ekman A,Pessa-Morikawa T,Iivanainen A
MA1-82041 was used in immunohistochemistry to evaluate a panel of antibodies and lectins for identifying the major cell types in paraffin sections of bovine tissues
Fri Jan 27 00:00:00 EST 2006
Cyclooxygenase-2 overexpression in MCF-10F human breast epithelial cells inhibits proliferation, apoptosis and differentiation, and causes partial transformation.
International journal of cancer
Lu S,Yu G,Zhu Y,Archer MC
MA1-82041 was used in western blot to investigate the effect of COX-2 overexpression on inhibiting proliferation, apoptosis and differentiation.
Mon Oct 10 00:00:00 EDT 2005
The absence of delayed graft function is predicted by the presence of manganese-superoxide dismutase in distal tubules of renal allografts.
Transplantation
Boom H,de Heer E,van der Wal A,Kruidenier L,de Fijter JW,Benediktsson H,Paul LC,van Es LA
MA1-82041 was used in immunohistochemistry to investigate the association between delayed graft function and manganese-superoxide dismutase in distal tubules of renal allografts
Wed Apr 27 00:00:00 EDT 2005
Sarcomatoid carcinoma of the renal pelvis with giant cell tumor-like features: case report with immunohistochemical findings.
International journal of urology : official journal of the Japanese Urological Association
Acikalin MF,Kabukcuoglu S,Can C
MA1-82041 was used in immunohistochemistry to report on a case of sarcomatoid carcinoma of the renal pelvis with giant cell tumor-like features
Tue Feb 01 00:00:00 EST 2005
Epithelioid angiomyolipoma of the kidney.
International journal of urology : official journal of the Japanese Urological Association
Acikalin MF,Tel N,Oner U,Pasaoglu O,Dönmez T
MA1-82041 was used in immunohistochemistry to report on a case of epithelioid angiomyolipoma of the kidney
Tue Feb 01 00:00:00 EST 2005
Pigmented solid-pseudopapillary neoplasm of the pancreas.
Pathology international
Daum O,Sima R,Mukensnabl P,Vanecek T,Brouckova M,Benes Z,Michal M
MA1-82041 was used in immunohistochemistry to report on a case of pigmented solid-pseudopapillary neoplasm of the pancreas
Sun May 01 00:00:00 EDT 2005
Mesenchymal hamartoma of the liver in adulthood: immunohistochemical profiles, clinical and histopathological features in two patients.
Journal of hepato-biliary-pancreatic surgery
Yesim G,Gupse T,Zafer U,Ahmet A
MA1-82041 was used in immunohistochemistry to study two adult patients with mesenchymal hamartoma of the liver
Thu Jul 06 00:00:00 EDT 2006
Oncocytic adrenocortical carcinomas: a pathological and immunohistochemical study of four cases in comparison with conventional adrenocortical carcinomas.
Pathology international
Song SY,Park S,Kim SR,Suh YL
MA1-82041 was used in immunohistochemistry - paraffin section to describe four cases of oncocytic adrenocortical carcinomas
Sun Aug 01 00:00:00 EDT 2004
Adrenocortical oncocytic tumors: report of 10 cases and review of the literature.
International journal of surgical pathology
Bisceglia M,Ludovico O,Di Mattia A,Ben-Dor D,Sandbank J,Pasquinelli G,Lau SK,Weiss LM
MA1-82041 was used in immunohistochemistry to report on 10 cases of adrenocortical oncocytic tumors
Thu Jul 01 00:00:00 EDT 2004
Recurrent amyloidoma of soft tissue with exuberant giant cell reaction.
Archives of pathology & laboratory medicine
Mukhopadhyay S,Damron TA,Valente AL
MA1-82041 was used in immunohistochemistry to report a clinical case of recurrent amyloidoma of soft tissue
Mon Dec 01 00:00:00 EST 2003
Automated acquisition of stained tissue microarrays for high-throughput evaluation of molecular targets.
The Journal of molecular diagnostics : JMD
Vrolijk H,Sloos W,Mesker W,Franken P,Fodde R,Morreau H,Tanke H
MA1-82041 was used in immunohistochemistry to study the use of an automated system for tissue microarray data acquisition and high throughput eveluation
Fri Aug 01 00:00:00 EDT 2003
Invasion of interstitial matrix by a novel cell line from primary peritoneal carcinosarcoma, and by established ovarian carcinoma cell lines: role of cell-matrix adhesion molecules, proteinases, and E-cadherin expression.
Gynecologic oncology
Kokenyesi R,Murray KP,Benshushan A,Huntley ED,Kao MS
MA1-82041 was used in western blot to study how cell-matrix interactions influence the invasive behavior of a novel, primary peritoneal carcinosarcoma cell line
Tue Apr 01 00:00:00 EST 2003
Ectopic thyroid in the gallbladder: report of a case.
Surgery today
Ihtiyar E,Isiksoy S,Algin C,Sahin A,Erkasap S,Yasar B
MA1-82041 was used in immunohistochemistry to describe an unusual case of ectopic thyroid tissue
Fri Feb 27 00:00:00 EST 2004