Applications | Tested Dilution | Publications |
---|---|---|
ELISA (ELISA) |
Assay-Dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
OTI2B2 |
Immunogen |
Full length human recombinant protein of human MAP2K1 produced in HEK293T cell. |
Conjugate |
Biotin
|
Form |
liquid |
Concentration |
0.5 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS with 10mg/mL BSA, 50% glycerol |
Contains |
0.05% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
MEK1/mitogen activated protein kinase/ERK kinase 1/microtubule-associated protein 2 kinase is a dual specificity kinase and catalyzes both a threonine and a tyrosine residue on MAP kinases ERK1 and ERK2. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines. Mutations in the gene can lead to cardiofaciocutaneous syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dual specificity mitogen-activated protein kinase kinase 1; EC 2.7.12.2; ERK activator kinase 1; kinase MEK1; MAP kinase kinase 1; MAPK/ERK kinase 1; MAPKK 1; MEK 1; MKK (Thr292); MKK (Thr386); MKK2; MP2K1; PRKMK2; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Gene Aliases: CFC3; MAP2K1; MAPKK1; MEK1; MKK1; PRKMK1
UniProt ID: (Human) Q02750
Entrez Gene ID: (Human) 5604
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support