Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For maximum recovery of product, centrifuge the vial prior to removing the cap.
ZPK is a member of serine/threonine protein kinase family. This kinase contains a leucine-zipper domain, and is predominately expressed in neuronal cells. The phosphorylation state of this kinase in synaptic terminals was shown to be regulated by membrane depolarization via calcineurin. This kinase forms heterodimers with leucine zipper containing transcription factors, such as cAMP responsive element binding protein (CREB) and MYC, and thus may play a regulatory role in PKA or retinoic acid induced neuronal differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DLK; Dual leucine zipper bearing kinase; dual leucine zipper kinase; Leucine-zipper protein kinase; M3KC; MAPK-upstream kinase; Mitogen activated protein kinase 12 (Zipper (leucine) protein kinase); mitogen activated protein kinase kinase kinase 12; Mitogen-activated protein kinase kinase kinase 12; Mixed lineage kinase; MUK; Zipper (leucine) protein kinase; ZPK
Gene Aliases: DLK; Map3k12; MUK; PK; Zpk
UniProt ID: (Rat) Q63796, (Mouse) Q60700
Entrez Gene ID: (Rat) 25579, (Mouse) 26404
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support