Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:1,000 | - |
Immunohistochemistry (IHC) |
1:50-1:250 | - |
ELISA (ELISA) |
1:10,000 | - |
Immunoprecipitation (IP) |
1:50-1:250 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Chicken / IgY |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Synthetic peptide taken within amino acid region 300-350 on human Mitogen-activated protein kinase 1. |
Conjugate |
Biotin |
Form |
Liquid |
Concentration |
0.5-1.5 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
proprietary buffer, pH 7.4-7.8, with 30% glycerol, 0.5% BSA |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
ERK2 (also known as MAPK1) is a member of the MAP kinase subfamily of Ser/Thr protein kinases. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERK-2; ERT1; Extracellular signal-regulated kinase 2; extracellular-signal-regulated kinase 2; MAP kinase 1; MAP kinase 2; MAP kinase isoform p42; MAPK 1; MAPK 2; mitogen activated protein kinase 1; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; p42-MAPK; protein tyrosine kinase ERK2
Gene Aliases: 9030612K14Rik; AA407128; AU018647; C78273; ERK; ERK-2; ERK2; ERT1; Mapk; MAPK1; MAPK2; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; PRKM1; PRKM2
UniProt ID: (Human) P28482, (Mouse) P63085, (Rat) P63086
Entrez Gene ID: (Human) 5594, (Mouse) 26413, (Rat) 116590
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support