Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
SLC25A51 gene ontology annotations related to this gene include transmembrane transporter activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitochondrial carrier triple repeat 1; mitochondrial carrier triple repeat protein 1; mitochondrial NAD(+) transporter SLC25A51; mitochondrial NAD+ transporter; unnamed protein product
Gene Aliases: 9130208E07Rik; CG7943; D130005A03Rik; Gm138; MCART1
UniProt ID: (Human) Q9H1U9, (Mouse) Q5HZI9
Entrez Gene ID: (Human) 92014, (Mouse) 230125
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support